Plastic General Purpose Burets
Thermo Scientific™ Nalgene™ Acrylic Burets with PMP Tip, TFE Stopcock and Plug Available on GSA/VA Contract for Federal Government customers only.
Accurately perform applications with Thermo Scientific™ Nalgene™ Acrylic Burets with PMP Tip, TFE Stopcock and Plug, which meet ASTM E287 standards.
PYREX™ Class A Buret with Product Standard PTFE Stopcock Plug Available on GSA/VA Contract for Federal Government customers only.
Buret has colored markings, fine, sharp lines and large easy-to-read numbers
PYREX™ Economy Grade Burets with Replaceable PTFE Stopcock Available on GSA/VA Contract for Federal Government customers only.
Replaceable components for cost savings
DWK Life Sciences Kimble™ KIMAX™ Schellbach Burets Available on GSA/VA Contract for Federal Government customers only.
With straight PTFE-plug No. 2 stopcocks and KimKap™ dust cap
PYREX™ Serialized/Certified Class A Burets Available on GSA/VA Contract for Federal Government customers only.
Colored markings, fine, sharp lines and large easy-to-read numbers
DWK Life Sciences Kimble™ KIMAX™ Buret Replacement Glass Barrels Available on GSA/VA Contract for Federal Government customers only.
Replacement glass barrels for Education-Grade Burets.
PYREX™ Serialized/Certified Class A Precision Bore Buret Available on GSA/VA Contract for Federal Government customers only.
Sharply defined and easy-to-read colored graduations
DWK Life Sciences Kimble™ KIMAX™ Schellbach Burets with Three-Way Stopcock Available on GSA/VA Contract for Federal Government customers only.
With No. 2 three-way stopcock with automatic PTFE plug
Fisherbrand™ Schellbach Autozeroing PTFE Buret, Three-Way Stopcock Available on GSA/VA Contract for Federal Government customers only.
Provides high quality at an affordable price.
EISCO Acrylic Buret Products for Science Education
Transparent acrylic body and leakproof PTFE plunger stopcock.
Supertek Scientific Acrylic Burets Products for Science Education
Transparent buret with leakproof stopcock fitted with self lubricating PTFE plug.
DWK Life Sciences Kimble™ ValueWare™ Buret Products for Science Education
Designed for use in educational institutions and for many general laboratory procedures.
GSC Go Science Crazy Glass Burette with PTFE Stopcock Products for Science Education
Avoid hassles by choosing a burette with a stopcock that won't freeze. The GSC International Glass Burette with PTFE Stopcock is subdivided by 0.1 ml, does not require grease and is nearly freeze proof.
  spinner