Metal Crucibles
Fisherbrand™ Zirconium Crucibles with Cover
Fisherbrand Zirconium Crucibles with Cover can withstand about 100 two- to five- minute fusions between 460°C (860°F) and 900°C (1652°F)
Fisherbrand™ Nickel Crucibles and Covers
Fisherbrand Nickel Crucibles can withstand temperatures up to 800°C (1472°F)
Fisherbrand™ Nickel Crucibles and Covers
Fisherbrand Nickel Crucibles can withstand temperatures up to 800°C (1472°F)
Alfa Aesar™ Graphite Fusion Crucible
Manufactured from graphite materials that provide exceptional thermal uniformity and exactness to dimensional specifications.
  spinner