Porcelain Crucibles
Fisherbrand™ High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
PYREX™ Gooch Type Filtering Crucibles Available on GSA/VA Contract for Federal Government customers only.
For drying precipitates to constant weight at 110°C (230°F)
DWK Life Sciences Kimble™ KIMAX™ Gooch Filtering Crucibles Available on GSA/VA Contract for Federal Government customers only.
Dry precipitates to constant weight at 110°C (230°F)
Fisherbrand™ Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
QSI Fused-Quartz Crucibles
Withstand temperatures up to 1250°C (2282°F)
Fisherbrand™ Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
CoorsTek™ High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
CoorsTek™ Porcelain Desiccator Plates for Crucibles
With 30mm diameter holes for crucibles
Fisherbrand™ Porcelain 90mL Dish Crucible
For laboratory and industrial purpose
Fisherbrand™ Porcelain Gooch Filtering Crucibles
For total suspended solids determinations
CoorsTek™ Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
CoorsTek™ Porcelain Coal Moisture Testing Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
Fisherbrand™ Porcelain Casserole Crucibles
For use in laboratory studies
Plasticoid Walter Crucible Holder
Rubber with removable glass stem ground at 45° angle
Plasticoid Crucible Holders
Holders for glass crucibles
Fisherbrand™ Porcelain Capsule-Form Crucibles
Glazed inside and out except for rim
QSI Fused Quartz Crucible Covers
For 08-072 Series Crucibles
CoorsTek™ CoorsTek™ Porcelain Crucibles with Filtering Discs
Disc permanently fused into crucible walls
Fisherbrand™ Porcelain Rose Crucible
For use in very high temperatures
CoorsTek™ Porcelain Gooch Filtering Crucibles
Glazed inside and out except for outside bottom surface
Fisherbrand™ Porcelain Bitumen Filtering Crucible
Filtration device for general laboratory use
Fisherbrand™ Porcelain Flat Capsule Crucibles
For use in laboratories of educational institutions, scientific development and industrial research, chemical, fertilizer, petro-chemical establishments
Fisherbrand™ Porcelain 25mL Crucibles
For laboratory and industrial purpose
GSC Go Science Crazy Crucible High Form with Lid, 50ml
The GSC International's High Form Crucible with Lid porcelain is uniform in thickness and dense which allows stable mechanical strength and little permeability. When properly used, the glazed pieces can withstand temperatures of 1050°C.
CoorsTek™ Porcelain Gooch Filtering Crucible for Extractions
Perforated bottom and two holes near rim
DWK Life Sciences DURAN™ Filter Crucible
Thermal shock and chemical resistant
  spinner