Cuvets and Cells
Thermo Scientific™ Spectrophotometer Test-tube Cuvettes
Thermo Scientific™ Test-Tube Cuvettes are easy-to-use, inexpensive spectrophotometer cells for routine analyses.
Thermo Scientific™ Electroporation Cuvettes
Increase the ease of sample identification and labelling with these premium-grade polycarbonate electroporation cuvettes, which have frosted, colored caps.
Fisherbrand™ Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators
Fisherbrand™ Disposable Cuvettes, Standard; Polystyrene
Choice of polystyrene or methacrylate
Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid
Fit standard spectrophotometer cell holders
Fisherbrand™ Round Cuvets Available on GSA/VA Contract for Federal Government customers only.
For use with commercial colorimeters and spectrophotometers.
Fisherbrand™ Absorption Cylindrical Cells
Fit standard spectrophotometer cell holders
TECHREF™ H-2358 Sample Cups/Cells, 48mm, single open ended
Designed specifically for use with X-ray fluorescence analyzers. TECHREF™ Sample Cells are available for XRF analyzers for a variety of styles and manufacturers and are designed for ease of use.
Fisherbrand™ Absorption Rectangular Quartz Flow Cells
Choose cells with quartz windows for transformation from the far ultraviolet through infrared
BrandTech™ BRAND™ UV-Cuvette Disposable Cuvets Available on GSA/VA Contract for Federal Government customers only.
Fit most spectrophotometers without adapters.
BTX™ Electroporation Chambers - Cuvettes Plus™
Field strengths up to 25kV/cm are possible when these cuvets are used with BTX generators
BrandTech™ Cuvet Caps Available on GSA/VA Contract for Federal Government customers only.
Round caps for Ultramicro UV-cuvettes
HemoCue America Glucose 201 and WBC Analyzer Microcuvettes
Serves as a sample container and reaction chamber
TECHREF™ C-1400 Sample Cups/Cells, 46mm, single open ended
Designed specifically for use with X-ray fluorescence analyzers. TECHREF™ Sample Cells are available for XRF analyzers for a variety of styles and manufacturers and are designed for ease of use.
Eppendorf™ UVette™ Cuvets Available on GSA/VA Contract for Federal Government customers only.
An alternative to the expensive, breakable quartz cuvets traditionally required for use at UV wavelengths
TECHREF™ C-2358 Sample Cups/Cells, 48mm, single open ended
Designed specifically for use with X-ray fluorescence analyzers. TECHREF™ Sample Cells are available for XRF analyzers for a variety of styles and manufacturers and are designed for ease of use.
Genlantis™ Zeus™ Electroporation Cuvets Non-distribution product as customer accommodation.
Compatible with all common electroporation devices
Fisherbrand™ Quartz Cuvette with PTFE Stopper
Contains quartz windows for transmission from far ultraviolet through the infrared.
TECHREF™ C-700 Sample Cups/Cells, 31 mm, single open ended
Designed specifically for use with X-ray fluorescence analyzers. TECHREF™ Sample Cells are available for XRF analyzers for a variety of styles and manufacturers and are designed for ease of use.
Greiner Bio-One Semi-micro/Macro Cuvettes
Ideal for enzymatic determinations. Greiner Bio-One Semi-micro/Macro Cuvettes are manufactured from crystal clear polystyrene.
  spinner