Separatory Funnels
Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ FEP with Closure Available on GSA/VA Contract for Federal Government customers only.
Utilize these chemically inert, translucent and autoclavable funnels which feature a TFE stopcock assembly. Chemical-resistant, leakproof and requires no lubrication.
Thermo Scientific™ Nalgene™ Polypropylene Separatory Funnels with Closure Available on GSA/VA Contract for Federal Government customers only.
Perform a variety of lab duties with chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Polypropylene Separatory Funnels.
Thermo Scientific™ Replacement Stopcock Assembly for Nalgene™ Separatory Funnels Available on GSA/VA Contract for Federal Government customers only.
Maintain separatory funnels with this Replacement Stopcock Assembly for Thermo Scientific™ Nalgene™ Separatory Funnels, which ensures a leakproof† fit without lubrication.
PYREX™ Cylindrical Separatory Funnel, Graduated, PTFE Standard Stopcock, Glass Standard Taper Stopper Available on GSA/VA Contract for Federal Government customers only.
Microfinish PTFE stopcock plugs reduce freezing problems and eliminate contaminating greases
DWK Life Sciences Kimble™ KIMAX™ Squibb Pear-Shaped Sep. Funnel with PTFE-Plug Stopcock, standard taper PE Stopper Available on GSA/VA Contract for Federal Government customers only.
Closed-bottom, linear high-density polyethylene stopper in standard taper ground neck
DWK Life Sciences Kimble™ Kontes™ Squibb Separatory Funnel Only Available on GSA/VA Contract for Federal Government customers only.
Replacement Funnel for KIMAX™ Squibb Separatory Funnel with PTFE Stopcock and Standard Taper Joint 636280-2000.
Fisherbrand™ Squibb Funnels with standard taper Stopcocks and Stoppers Available on GSA/VA Contract for Federal Government customers only.
Range of separatory funnels manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
PYREX™ Squibb Separatory Funnel, PTFE Product Standard Stopcock, Standard Taper Joints Available on GSA/VA Contract for Federal Government customers only.
With No. 2 Product Standard stopcocks and inner and outer 19/22 Standard Taper joints.
PYREX™ Squibb Glass Separatory Funnels with Screw Cap and PTFE Valve Available on GSA/VA Contract for Federal Government customers only.
Constructed of durable PYREX borosilicate glass
DWK Life Sciences Kimble™ KIMAX™ Squibb Funnels with standard taper Stopcock and Stopper Available on GSA/VA Contract for Federal Government customers only.
Lower stems made of large-diameter tubing for liquid to drain out completely when stopcock is closed
PYREXPLUS™ Coated Squibb Separatory Funnel, PTFE Standard Stopcock, Glass Standard Taper Stopper Available on GSA/VA Contract for Federal Government customers only.
Protective PVC coating reduces spills and helps prevent glass from shattering
Eisco 250ml Separating Funnel - Borosilicate Glass, Pear Shaped, With Stopper And Rota flow Stopcock
250ml Separating Funnel - Borosilicate Glass, Pear Shaped, with Stopper and Rota flow Stopcock
Eisco 1000ml Separating Funnel - Pear Shaped Borosilicate Glass - Rota flow Stopcock
Borosilicate glass with interchangeable plastic stopper and screw type Rota flow stopcock. Internal diameter of top opening is 1.25 in. Cap. 1000 ml, Socket size 29/32
PYREX™ Squibb Pear-Shaped Separatory Funnels with Rotaflo PTFE-Plug Stopcock Available on GSA/VA Contract for Federal Government customers only.
Pear-shaped with standard taper ground neck, offset PTFE-plug stopcock and all-glass PYREX stopper
Synthware™ Squibb Separatory Funnels with Glass Stopcock
Supplied complete with flask length glass stopper
PYREX™ Economy Squibb Separatory Funnel, Polyethylene Standard Taper Stopper, Replaceable Standard PTFE Stopcock Available on GSA/VA Contract for Federal Government customers only.
Economical separatory funnel designed to reduce replacement costs by providing replaceable components
  spinner