Filling Funnels
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels Available on GSA/VA Contract for Federal Government customers only.
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels, featuring long stems and external ribbing that eliminates air lock.
PYREX™ Seamless Funnels Available on GSA/VA Contract for Federal Government customers only.
Bottoms are ground flat
Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels Edge Available on GSA/VA Contract for Federal Government customers only.
Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.
PYREX™ Funnel for Sugar Analysis Available on GSA/VA Contract for Federal Government customers only.
Molded construction with bottom ground flat and beaded rim
DWK Life Sciences Kimble™ KIMAX™ Filling Funnels Available on GSA/VA Contract for Federal Government customers only.
Fire-polished edges reduce chipping
PYREX™ Powder Funnel Edge Available on GSA/VA Contract for Federal Government customers only.
With short wide stems
Fisherbrand™ Powder Funnels Edge Available on GSA/VA Contract for Federal Government customers only.
Parallel stems minimize bridging of powder
Fisherbrand™ Reusable Glass Short-Stem Funnels Available on GSA/VA Contract for Federal Government customers only.
High resistance to chemical attack, mechanical, and thermal shock
PYREX™ 60° Angle Fluted Funnel, Short Stem Available on GSA/VA Contract for Federal Government customers only.
Useful in clinical and hospital labs; can be used in flasks or tubes without racks
Bel-Art™ SP Scienceware™ Polypropylene Powder Funnels Available on GSA/VA Contract for Federal Government customers only.
Feature a large bore stem to minimize bridging of powder
Bel-Art™ SP Scienceware™ Polypropylene Funnels with Short Stems and External Ridges
Sturdy, polypropylene funnels with ridges help prevent vapor lock
Thermo Scientific™ Nalgene™ Large HDPE Funnels Available on GSA/VA Contract for Federal Government customers only.
Choose a safer alternative to glass with the shatterproof Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release.
Fisherbrand™ Long-Stem Analytical Funnels Available on GSA/VA Contract for Federal Government customers only.
Molded polypropylene with external ridges and outside ribs to prevent vapor lock
DWK Life Sciences Kimble™ KIMAX™ Long Stem 60° Angle Addition Funnels Available on GSA/VA Contract for Federal Government customers only.
High resistance to chemical attack, mechanical and thermal shock
Bel-Art™ SP Scienceware™ Utility Funnels Available on GSA/VA Contract for Federal Government customers only.
Externally ribbed, polypropylene funnels suitable for a variety of general lab applications
Caplugs™ (Evergreen Scientific) Disposable Funnels
Economical funnels for a variety of applications.
Thermo Scientific™ Nalgene™ Utility Funnels Available on GSA/VA Contract for Federal Government customers only.
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels, which are an excellent shatter-proof alternative.
Fisherbrand™ Heavy-Duty/Utility Funnels Available on GSA/VA Contract for Federal Government customers only.
Ideal for continuous lab work and heavy-duty jobs
Bel-Art™ SP Scienceware™ Polypropylene Long-Stemmed Funnels Available on GSA/VA Contract for Federal Government customers only.
Heavy-duty design with external ribs to prevent air lock
PYREX™ 60° Angle Fluted Funnel with Long Stem Available on GSA/VA Contract for Federal Government customers only.
Depressed inside flutings increase filtering speed
Bel-Art™ SP Scienceware™ Large Powder Funnel Available on GSA/VA Contract for Federal Government customers only.
For transfer of powders and general use
Variety Glass Variety Glass Mooney Air Vent for Liquids
Heavy-molded flint glass funnels with smooth rims, inside ribs
Thermo Scientific™ Nalgene™ Heavy-Duty LDPE Funnels Available on GSA/VA Contract for Federal Government customers only.
Avoid breakage and corrosion with Thermo Scientific™ Nalgene™ Heavy-Duty LDPE Funnels, the toughest large funnels available.
Fisherbrand™ Reusable Glass Long-Stem Funnels Available on GSA/VA Contract for Federal Government customers only.
Promote faster, more effective filtering
PYREX™ Plain 60° Angle Funnel with Long Narrow Stem Available on GSA/VA Contract for Federal Government customers only.
Permit precise filter paper fitting
  spinner