Special Purpose Funnels
Fisherbrand™ Glass Funnel Available on GSA/VA Contract for Federal Government customers only.
Provides for easy buret filling.
Simport™ Scientific Disposable Funnel
Tapered at a 60° angle with inside fluting
Justrite™ Large Steel Drum Funnel With Self-Closing Cover Available on GSA/VA Contract for Federal Government customers only.
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Large Steel Drum Funnel With Self-Closing Cover helps to prevent spills when transferring flammable solvents, thinners, oils, or paint wastes. This self-closing model meets NFPA1 and the International Fire Code. It shuts and locks automatically when a fusible link melts at 165°F (74°C) under fire conditions.
Heidolph™ Glassware - Dropping Funnels Available on GSA/VA Contract for Federal Government customers only.
For use with Heidolph reactors and evaporators
  spinner