Plastic Filling Funnels
Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels Available on GSA/VA Contract for Federal Government customers only.
Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels Available on GSA/VA Contract for Federal Government customers only.
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels, featuring long stems and external ribbing that eliminates air lock.
Fisherbrand™ Powder Funnels Available on GSA/VA Contract for Federal Government customers only.
Parallel stems minimize bridging of powder
Thermo Scientific™ Nalgene™ Heavy-Duty LDPE Funnels Available on GSA/VA Contract for Federal Government customers only.
Avoid breakage and corrosion with Thermo Scientific™ Nalgene™ Heavy-Duty LDPE Funnels, the toughest large funnels available.
Thermo Scientific™ Nalgene™ Utility Funnels Available on GSA/VA Contract for Federal Government customers only.
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels, which are an excellent shatter-proof alternative.
Fisherbrand™ Heavy-Duty/Utility Funnels Available on GSA/VA Contract for Federal Government customers only.
Ideal for continuous lab work and heavy-duty jobs
Caplugs™ Disposable Funnels
Economical funnels for a variety of applications.
Fisherbrand™ Long-Stem Analytical Funnels Available on GSA/VA Contract for Federal Government customers only.
Molded polypropylene with external ridges and outside ribs to prevent vapor lock
Fisherbrand™ Large LDPE Funnels
Large plastic funnels for transferring liquids at temperatures up to 80°C (180°F).
Bel-Art™ SP Scienceware™ Polypropylene Long-Stemmed Funnels Available on GSA/VA Contract for Federal Government customers only.
Heavy-duty design with external ribs to prevent air lock
Thermo Scientific™ Nalgene™ Large HDPE Funnels Available on GSA/VA Contract for Federal Government customers only.
Choose a safer alternative to glass with the shatterproof Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release.
Bel-Art™ SP Scienceware™ Utility Funnels Available on GSA/VA Contract for Federal Government customers only.
Externally ribbed, polypropylene funnels suitable for a variety of general lab applications
Bel-Art™ SP Scienceware™ Polypropylene Funnels with Short Stems and External Ridges
Sturdy, polypropylene funnels with ridges help prevent vapor lock
Bel-Art™ SP Scienceware™ Polypropylene Powder Funnels Available on GSA/VA Contract for Federal Government customers only.
Feature a large bore stem to minimize bridging of powder
Bel-Art™ SP Scienceware™ Large Powder Funnel Available on GSA/VA Contract for Federal Government customers only.
For transfer of powders and general use
Bel-Art™ SP Scienceware™ Urbanti™ High-Speed Funnels Available on GSA/VA Contract for Federal Government customers only.
Optimized for nonvacuum, high-speed filtration, these glass-clear funnels have internal helicoid ribs to increase the filtration speed
Eisco Funnel, Dia 2 1/4 in. with a 1/4 in. outlet and measures 3 1/2 in. tall
This plastic funnel is chemically resistant, durable, and easy to clean.
Bel-Art™ SP Scienceware™ Micro Funnels Available on GSA/VA Contract for Federal Government customers only.
Designed for micro work, filling burets, manometers and small bottles
Eisco Funnel, Dia 3 15/16 in. with a 3/8 in. outlet and measures 6 in. tall
This plastic funnel is chemically resistant, durable, and easy to clean.
Eisco Funnel, Dia 2 3/4 in. with a 1/4 in. outlet and measures 4 1/2 in. tall
This plastic funnel is chemically resistant, durable, and easy to clean.
Bel-Art™ SP Scienceware™ Standard Stem Funnels Available on GSA/VA Contract for Federal Government customers only.
Feature an exclusive filter-step internal design for speedier filtrations
Eisco Funnel, Dia 3 1/4 in. with a 1/4 in. outlet and measures 5 1/2 in. tall
This plastic funnel is chemically resistant, durable, and easy to clean.
Eisco Funnel, Dia 3 15/16 in. with a 3/8 in. outlet and measures 6 in. tall
This plastic funnel is chemically resistant, durable, and easy to clean.
Bel-Art™ SP Scienceware™ Large Industrial Drum Funnel Available on GSA/VA Contract for Federal Government customers only.
Allows the user to safely fill drums and containers
  spinner