Condensers
DWK Life Sciences Kimble™ KIMAX™ West Condensers Available on GSA/VA Contract for Federal Government customers only.
With sloped inner tube and small annular space
PYREX™ Cold Finger Condenser With 24/40 Inner Joints
Used to regulate refluxing by adjusting flow of air or water
DWK Life Sciences Kimble™ Kontes™ West Condensers with Full Length 24/40 Joints
Condenser with a standard outer joint at the top and a standard taper inner drip joint at the bottom
PYREX™ Replacement Allihn Condenser
Replacement Allihn condenser with bulbs
DWK Life Sciences Kimble™ Liebig Medium Length Condenser with 14/20 Joints
Design eliminates the use of bulbs where holdup is likely to occur.
Synthware™ Liebig Condenser
With standard taper outer joint at the top and lower inner drip tip joint
Synthware™ Reflux Condenser, Jacketed
Jacketed reflux condenser has I.D. of inner tube large enough to accept a thermometer
PYREX™ Air Condensers With Low Actinic Glass
Low actinic glass protects light sensitive materials
Synthware™ West Condensers
Narrow annular space for fast coolant speed, provides high cooling efficiency
Synthware™ Allihn Condenser
With joints at the top and the bottom
DWK Life Sciences Kimble™ Kontes™ Liebig Condensers
Standard Taper outer joint at the top; standard Taper inner drip joint at the bottom.
DWK Life Sciences Kimble™ Kontes™ Allihn Condensers with Straight Outlet Tube
For refluxing applications including Soxhlet extractions
Synthware™ Allihn Condenser for Soxhlet Extractor
For use with Soxhlet extraction apparatus
Synthware™ Air Reflux Column, 14/20 Joints, Microscale
Condenser uses surrounding air to dissipate heat during refluxing
Synthware™ Micro Dewar Condenser
Large opening in the top for use with dry ice/acetone
Synthware™ Air Reflux Condenser, Microscale
Uses surrounding air to dissipate heat during refluxing
Synthware™ Cold Finger Condensers, with Drip Tip
Inner joint and a sealed extended lower tip for drop control
Synthware™ Liebig Condenser, with Removable Hose Connections
Condenser has a standard taper outer joint at the top and a lower inner drip tip joint
Synthware™ Coiled Condensers
With an inner coil that coolant is circulated through
Synthware™ Reflux Condenser
For use with volatile solvents
DWK Life Sciences Kimble™ Kontes™ Coil Type Reflux Condenser with Removable Hose Connections
Detachable polypropylene hose connections provide a safe and easy way to connect and disconnect tubing.
PYREX™ West Condensers with Drip Tip
For general condensation applications
  spinner