Glass Specialty Rotary Traps
Labconco™ Activated Carbon Solvent Trap for Rotary Vane Vacuum Pumps
Activated Carbon Solvent Trap protects a rotary vane vacuum pump from organic solvent vapors.
Synthware™ Rotary Evaporator Anti-Splash Adapters, Modified
With two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask
Fisherbrand™ Rotary Evaporator Trap with Drain Holes
Use where foaming is likely to occur and be drawn into the condenser.
Synthware™ Rotary Evaporator Anti-Splash Adapters
Prevents possible contamination from rotary evaporator
DWK Life Sciences Kimble™ Kontes™ Rotary Evaporator Traps With Inner Vapor Tube at Top
Design allows for expanded volume for mixtures that have a tendency to foam excessively
PYREX™ Evaporator Traps Available on GSA/VA Contract for Federal Government customers only.
Used when foam is likely to be created and drawn into the condenser of rotary evaporator
Synthware™ Distilling Trap Adapters
Placed above flask to prevent carry-over of reactions which have a tendency to foam
Fisherbrand™ Kontes™ 100mL Rotary Evaporator Trap with Square Joint Bead, with Drain Holes Available on GSA/VA Contract for Federal Government customers only.
Use where foaming is likely to occur and be drawn into the condenser. Available in 160mm length with 14/20 bottom joints or 180mm length with 24/40 bottom joints.
Synthware™ Anti-Climb Adapters
Trap provides extended volume for reactions that have tendency to foam excessively
DWK Life Sciences Kimble™ Kontes™ Self Washing with Square Joint Bead Rotary Evaporator Trap
This trap is designed to eliminate hold-up common to the round style of rotary evaporator traps. Available in 100mL or 250mL capacities, with 14/20 or 24/40 bottom taper joints and 24/40 top taper joints.
DWK Life Sciences Kimble™ Kontes™ Fritted Rotary Evaporator Bump Trap
This trap has a 40-60 micron porosity fritted disc sealed between the lower inner and top outer joint to prevent the contents of the flask from entering into the condenser in the event of bumping.
DWK Life Sciences Kimble™ Kontes™ Rotary Evaporator Traps With Inner Vapor Tube at Bottom Available on GSA/VA Contract for Federal Government customers only.
Use when foam is likely to be created and drawn into the condenser
  spinner