Glass Centrifuge Bottles
Kimble™ KIMAX™ Babcock Bottles for Cheese Testing Available on GSA/VA Contract for Federal Government customers only.
Paley centrifuge bottles feature large stoppered opening next to neck to accommodate both liquids and solids
Kimble™ KIMAX™ Babcock Bottles for Ice Cream Testing Available on GSA/VA Contract for Federal Government customers only.
Scale is marked with permanent brown stain on neck.
Kimble™ KIMAX™ Brand Babcock Bottle for Skim Milk Testing Available on GSA/VA Contract for Federal Government customers only.
Graduated neck with narrow bore to fill through
PYREX™ Glass Centrifuge Bottles Available on GSA/VA Contract for Federal Government customers only.
Permit concentration of sediment for decanting operations
Kimble™ KIMAX™ Babcock Bottle for Gasoline Testing Available on GSA/VA Contract for Federal Government customers only.
For determination of unsaturated hydrocarbons in gasoline
Kimble™ KIMAX™ Babcock Bottles for Milk Testing Available on GSA/VA Contract for Federal Government customers only.
8% Bottle with “Sealed 3” marking
  spinner