Plastic Dispensing Bottles
Thermo Scientific™ Nalgene™ LDPE Drop-Dispensing Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Conveniently operate translucent Thermo Scientific™ Nalgene™ LDPE Drop-Dispensing Bottles with Closure one-handed; bottles feature tethered spout caps which snap securely in place.
Thermo Scientific™ Nalgene™ PPCO Dispensing Bottle with Closure: Autoclavable Available on GSA/VA Contract for Federal Government customers only.
Dispense sterile solutions with these PPCO dispensing bottles. The tethered spout cap snaps securely into place and won't pop off or leak when the closed bottle is squeezed.
Thermo Scientific™ Nalgene™ Teflon™ FEP Drop-Dispensing Bottle with Dropping Closure and Cap
Dispense strong acids/alkalis, reagents and solvents with these FEP drop-dispensing bottles which feature excellent chemical resistance for dispensing virtually any chemical.
Qorpak™ Flip Top Dispensing Bottles Available on GSA/VA Contract for Federal Government customers only.
For dispensing small amounts of liquid
Bel-Art™ SP Scienceware™ Measure-Matic Dispenser Available on GSA/VA Contract for Federal Government customers only.
Squeezing forces the desired volume, 5mL to 25mL, of liquid through the tube and into the cup
Fisherbrand™ LDPE Dispensing Bottles
Versatile, reliable bottles and spouts
Wheaton™ LDPE Dropping Bottles With Streaming Tips and Polypropylene Caps Available on GSA/VA Contract for Federal Government customers only.
Tip dispenses a stream of liquid or noncontrolled drop
Wheaton™ LDPE Dropping Bottles Available on GSA/VA Contract for Federal Government customers only.
Excellent chemical resistance
Bel-Art™ SP Scienceware™ Dispensing/Drop Bottles Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Easy-squeeze bottles for critical drop control.
Qorpak™ Trigger Spray Bottles Available on GSA/VA Contract for Federal Government customers only.
Spray pattern from fine mist to coarse spray to stream
Bel-Art™ SP Scienceware™ Dispensing/Drop Bottles Available on GSA/VA Contract for Federal Government customers only.
Easy-squeeze dispensing bottles allow for critical drop control
Bel-Art™ SP Scienceware™ Indicator Bottles Available on GSA/VA Contract for Federal Government customers only.
Double-duty bottles offer two ways to assist in staining slides
Qorpak™ Industrial Trigger Spray Bottle
Reduces product permeation during dipensing
Fisherbrand™ PTFE Dispensing Bottle
Heavy wall, wide mouth PTFE screw closure with machine finish for ease in cleaning
Qorpak™ Yorker Top Bottles with Red Tip Available on GSA/VA Contract for Federal Government customers only.
Offers a degree of light protection
Qorpak™ Nasal Spray Bottle
Made of squeezable and resilient LDPE
GIBCO™ Bottle Weight Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Keeps bottles upright in water baths to reduce risk of contamination
Dynalon™ Dispensing Bottles Products for Science Education
Low-density polyethylene bottles.
Dynalon™ Large Dispenser Bottle Products for Science Education
Standard plunger dispenses 1.01 oz. (30mL) per stroke.
  spinner