Environmental Sampling Bottles
Thermo Scientific™ Wide-Mouth Tall-Profile Clear Glass Jars with Closure
Designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ Wide-Mouth Short-Profile Clear Glass Jars with Closure
Designed for collecting soil, sludge and waste samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is required.
Thermo Scientific™ Bulk Pack HDPE Cylinder Rounds without Caps
HDPE Cylinder Round is a general-purpose container. Bulk and unassembled. Unprocessed, ready for your own cleaning procedure and assembly with caps purchased separately.
Thermo Scientific™ Wide-Mouth Amber Glass Packer with Closure
Designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required.
Thermo Scientific™ HDPE Cylinder Round Bottles
Collect water samples and other low-viscosity liquids to be analyzed for inorganic trace analytes with the original EPA-specified bottle for liquid sampling.
Thermo Scientific™ Narrow-Mouth Glass Septa Bottles with Open-Top Closure
These narrow-mouth type III soda-lime glass septa bottles are available as clear or amber, and are recommended for EPA Volatile Organic Analysis (VOA) Methods.
Thermo Scientific™ Wide-Mouth HDPE Packers with Closure
Recommended for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes.
Thermo Scientific™ Wide-Mouth Septa Jars
These wide-mouth type III soda-lime glass septa jars are available as clear or amber, and are recommended for EPA Volatile Organic Analysis (VOA) soil methods.
Thermo Scientific™ Boston Round Narrow-Mouth Amber Jugs with Closure
These “certified” containers meet or exceed EPA standards for semi-volatiles, pesticides, PCB«s and metals analyses.
Thermo Scientific™ LDPE Foam-lined Polypropylene Caps for Bulk Separates
LDPE-Lined White Caps are for general-purpose HDPE bulk separate containers.
Thermo Scientific™ Nalgene™ Certified Wide-Mouth HDPE Bottle with Polypropylene Screw Closure
Recommended for critical projects where sample containment and transport is of elevated concern, featuring durable construction that withstands the rigors of field operations.
Thermo Scientific™ PTFE-Lined Closed Caps
PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
Thermo Scientific™ Color-coded Sample Alert Labels
Increase the safety and efficiency of sample handling and identification with Thermo Scientific™ Color-Coded Sample Alert Labels.
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle with Closure
Nalgene Wide-Mouth HDPE Sterile Sample Bottles are break-resistant, guaranteed leak-proof and certified sterile for water sample collection for microbiological analyses.
Thermo Scientific™ Wide-Mouth VOA Glass Jars with Closure
Designed for collecting soil samples for VOA testing using methods where a pierceable cap is not required. Amber jars are recommended for light-sensitive analytes.
Thermo Scientific™ LDPE Cubitainers™ with LDPE Foam-lined Closure
Light weight,space-efficient solution for sampling in remote locations. Available certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride.
Thermo Scientific™ Wide-Mouth HDPE Oblong with Lined Closure
Save valuable space and stabilize samples within the shipping container. Certified to meet EPA Performance Based Specifications for metals, cyanide and fluoride analyses.
Thermo Scientific™ Loose Septa for 24-414 Open Top Caps
Thermo Scientific™ Loose Septa are replacement septa for Thermo Scientific VOA vials and septa bottles with 24-414 open top caps.
Thermo Scientific™ Security-Snap™ Coliform Polypropylene Water Sample Bottle: Sterile
Security-Snap Sterile Coliform Water Sample Bottle meets all the EPA requirements for microbiology sample containers for the analysis of drinking water.
Thermo Scientific™ Wide-Mouth HDPE Packers without Caps: Bulk Pack
HDPE Wide-Mouth Packers are general-purpose containers, bulk and unassembled. Unprocessed, ready for your own cleaning procedure and assembly with caps purchased separately.
Thermo Scientific™ Wide-Mouth Short-Profile Amber Glass Jars with Closure
Designed for collecting soil, sludge and waste samples for a variety of organic and inorganic target analytes where light protection is required.
Thermo Scientific™ Particle Certified HDPE Containers
Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.
Thermo Scientific™ Screw-Top Sterile Coliform Water Sample Bottle: Polystyrene, Polyethylene and Polypropylene
Designed for use with EPA compliant total coliform and fecal coliform water analysis using the Colilert analysis method or equivalent.
Thermo Scientific™ Wide-Mouth HDPE Packers with Closure, 1000mL, unprocessed
Recommended for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes.
Thermo Scientific™ HDPE Jugs with White LDPE Foam-Lined Polypropylene Screw Closure
Thermo Scientific™ HDPE Jugs meet or exceed EPA performance-based standards for metals, cyanide and fluoride.
Thermo Scientific™ Boston Round Narrow-Mouth Clear Glass Bottles with Closure
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ Straight-Sided Wide-Mouth HDPE Jar with Closure
Wide-Mouth HDPE Jar allows for easy filling and sample retrieval, and is Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride.
Thermo Scientific™ 5G Terra Core Soil Sampler
Save time with this no fuss, no mess disposable transfer tool for soil sampling.
  spinner