Glass Environmental Sampling Bottles
Thermo Scientific™ Wide-Mouth Tall-Profile Clear Glass Jars with Closure
Designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ Wide-Mouth Short-Profile Amber Glass Jars with Closure
Designed for collecting soil, sludge and waste samples for a variety of organic and inorganic target analytes where light protection is required.
Thermo Scientific™ Wide-Mouth Short-Profile Clear Glass Jars with Closure
Designed for collecting soil, sludge and waste samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ Wide-Mouth Amber Glass Packer with Closure
Designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required.
Thermo Scientific™ Wide-Mouth VOA Glass Jars with Closure
Designed for collecting soil samples for VOA testing using methods where a pierceable cap is not required. Amber jars are recommended for light-sensitive analytes.
Thermo Scientific™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is required.
Thermo Scientific™ Boston Round Narrow-Mouth Clear Glass Bottles with Closure
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ Boston Round Narrow-Mouth Amber Jugs with Closure
These “certified” containers meet or exceed EPA standards for semi-volatiles, pesticides, PCB«s and metals analyses.
Thermo Scientific™ Wide-Mouth Septa Jars
These wide-mouth type III soda-lime glass septa jars are available as clear or amber, and are recommended for EPA Volatile Organic Analysis (VOA) soil methods.
Thermo Scientific™ Depyrogenated Glass Containers
Save on expensive capital equipment investments with ready-to-use glassware in a wide variety of sizes and configurations.
Thermo Scientific™ Narrow-Mouth Glass Septa Bottles with Open-Top Closure
These narrow-mouth type III soda-lime glass septa bottles are available as clear or amber, and are recommended for EPA Volatile Organic Analysis (VOA) Methods.
Thermo Scientific™ Narrow-Mouth Closed Top VOA Bottles
Conduct EPA Volatile Organic Analysis (VOA) analysis using these narrow-mouth glass bottles featuring closed caps, for VOA testing where a pierceable closure is not required.
Thermo Scientific™ Particle Certified Glass Containers
Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Fisherbrand™ Certified Cleaned Amber Boston Round Bottles
Ideal storage for solvents, chemicals and samples. Fisherbrand™ Certified Cleaned Amber Boston Round Bottles are manufactured from amber colored soda glass.
Fisherbrand™ Custom Cleaned Amber Wide Mouth Packer Bottles
Ideal storage for light sensitive dry powders. Fisherbrand™ Custom Cleaned Amber Wide Mouth Packer Bottles are manufactured using amber soda glass.
Fisherbrand™ Certified Low-Particulate Boston Round Bottles
Ideal storage for solvents, chemicals and samples. Fisherbrand™ Certified Low Particulate Boston Round Bottles are manufactured from clear soda glass.
Fisherbrand™ Custom Cleaned Clear Boston Round Bottles
Ideal storage for solvents, chemicals and samples. Fisherbrand™ Custom Cleaned Clear Boston Round Bottles are manufactured from clear soda glass.
Fisherbrand™ Certified Cleaned Amber Wide Mouth Packer Bottles
Ideal storage for light sensitive dry powders. Fisherbrand™ Certified Cleaned Amber Wide Mouth Packer Bottles are manufactured from amber colored soda glass.
Fisherbrand™ Certified Low Particulate Wide Mouth Packer Bottles
Ideal for the storage of dry powders. Fisherbrand™ Certified Low Particulate Wide Mouth Packer Bottles are manufactued from clear soda glass.
Fisherbrand™ Custom Cleaned Clear Boston Rounds
Containers and caps are noncertified and precleaned with USP purified water
Fisherbrand™ Custom Cleaned Amber Boston Round Bottles
Ideal storage for solvents, chemicals and samples. Fisherbrand™ Custom Cleaned Amber Boston Round Bottles are manufactured from amber colored soda glass.
Fisherbrand™ Certified Low Particulate Wide Mouth Packer Bottles
Ideal for the storage of dry powders. Fisherbrand™ Certified Low Particulate Wide Mouth Packer Bottles are manufactured from clear soda glass.
Thermo Scientific™ Specialty Waters
Manufactured, packaged and tested to exacting standards, these specialty waters are highly purified low TOC water in 1L bottles or 4L glass jugs.
  spinner