Glass Bottles
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles Available on GSA/VA Contract for Federal Government customers only.
Store dry or liquid materials in Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles, ideal as inner containers in combination packaging.
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Store and ship liquid reagents with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure.
Thermo Scientific™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is required.
Thermo Scientific™ Wide-Mouth Tall-Profile Clear Glass Jars with Closure
Designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles Available on GSA/VA Contract for Federal Government customers only.
Utilize these safe and economical bottles as inner containers for designing, assembling and certifying combination packaging.
Fisherbrand™ Square Specimen Bottles Available on GSA/VA Contract for Federal Government customers only.
Perfect for storing and transporting specimens.
Thermo Scientific™ Open-Top Septa Caps, 24-414mm, bonded septa, bulk
Thermo Scientific™ Septa Caps include a bonded-in 0.045″ thick septa.
Fisherbrand™ Labeled Wash Bottles Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Polyethylene bottles with polypropylene closures to ensure leakproof dispensing.
Thermo Scientific™ Wide-Mouth Short-Profile Clear Glass Jars with Closure
Designed for collecting soil, sludge and waste samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ Boston Round Narrow-Mouth Clear Glass Bottles with Closure
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is not required.
Thermo Scientific™ PTFE-Lined Closed Caps
PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
Thermo Scientific™ Loose Septa for 24-414 Open Top Caps
Thermo Scientific™ Loose Septa are replacement septa for Thermo Scientific VOA vials and septa bottles with 24-414 open top caps.
Thermo Scientific™ Wide-Mouth Septa Jars
These wide-mouth type III soda-lime glass septa jars are available as clear or amber, and are recommended for EPA Volatile Organic Analysis (VOA) soil methods.
Thermo Scientific™ 5G Terra Core Soil Sampler
Save time with this no fuss, no mess disposable transfer tool for soil sampling.
Thermo Scientific™ Wide-Mouth VOA Glass Jars with Closure
Designed for collecting soil samples for VOA testing using methods where a pierceable cap is not required. Amber jars are recommended for light-sensitive analytes.
Thermo Scientific™ Narrow-Mouth Glass Septa Bottles with Open-Top Closure
These narrow-mouth type III soda-lime glass septa bottles are available as clear or amber, and are recommended for EPA Volatile Organic Analysis (VOA) Methods.
Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Prevent separation when squeezing Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure.
Thermo Scientific™ Color-coded Sample Alert Labels
Increase the safety and efficiency of sample handling and identification with Thermo Scientific™ Color-Coded Sample Alert Labels.
Thermo Scientific™ Wide-Mouth Amber Glass Packer with Closure
Designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required.
Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.
Thermo Scientific™ Heat-Sealable Polyethylene Bags
Protect specimens for mailing with Thermo Scientific™ Heat-Sealable Polyethylene Bags.
Thermo Scientific™ Nalgene™ Unitary™ Red LDPE Safety Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Easily identify Thermo Scientific™ Nalgene™ Unitary™ Red LDPE Safety Wash Bottles, featuring a wide mouth for faster, safe filling.
Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Dispense contents without tipping or shaking by using Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles, designed with a wide mouth for faster, easier filling.
Thermo Scientific™ LDPE Foam-lined Polypropylene Caps for Bulk Separates
LDPE-Lined White Caps are for general-purpose HDPE bulk separate containers.
Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Simplify content identification with these narrow-mouth wash bottles that feature colored closures with an integrated stem.
Thermo Scientific™ Slide Mailer and Folder Available on GSA/VA Contract for Federal Government customers only.
Hold up to 20 slides with the Thermo Scientific™ Slide Mailer and Folder.
  spinner