Plastic Bottles
Thermo Scientific™ Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Easily measure and dispense using these translucent, square narrow-mouth PPCO bottles, featuring mouths wider than conventional narrow-mouth bottles.
Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles, which are resistant to most acids, bases and alcohols.
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Store, ship and package liquids with these highly reliable, lab quality bottles, specifically designed for long-term demanding lab use.
Thermo Scientific™ Nalgene™ Square Polycarbonate Graduated Bottles without Closure: Tray Pack Available on GSA/VA Contract for Federal Government customers only.
These bottles are durable and autoclavable, and feature an easy-to-handle square design. They are ideal for storage of aqueous solutions and short-term storage of media.
Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Carry out applications such as trace metal analysis with flexible, impact-and chemical-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure.
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Store and ship liquid reagents with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure.
Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Save space with the rectangular design of Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure, a good choice when space is at a premium.
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays Available on GSA/VA Contract for Federal Government customers only.
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile media bottles, ideal for prolonged storage of pH sensitive liquids.
Thermo Scientific™ LDPE Cubitainers™ with LDPE Foam-lined Closure
Light weight,space-efficient solution for sampling in remote locations. Available certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride.
Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Ensure continued stock of Thermo Scientific™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure with this bulk-packed option.
Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Store samples and ship small volumes of valuable reagents and light sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure.
Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles Available on GSA/VA Contract for Federal Government customers only.
Store, ship and package light-sensitive liquid using these narrow-mouth, amber HDPE bottles. They offer excellent chemical resistance to most acids, bases and alcohols.
Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Freeze samples effectively with impact-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure.
Thermo Scientific™ Nalgene™ Square Polycarbonate Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Store aqueous solutions with durable and autoclavable Thermo Scientific™ Nalgene™ Square Polycarbonate Bottles, also ideal for short-term storage of media.
Thermo Scientific™ Nalgene™ Square PET Media Bottles without Closure: Sterile, Shrink-Wrapped Trays Available on GSA/VA Contract for Federal Government customers only.
Nalgene Square PET Media Bottles are sterile, lightweight and shatter-resistant.
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Eliminate costly washing, depyrogenation and autoclaving steps with these bottles, ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.
Thermo Scientific™ Wide-Mouth HDPE Packers with Closure
Recommended for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes.
Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles Available on GSA/VA Contract for Federal Government customers only.
Eliminate the cost of cleaning, sterilizing and associated validations when compared to bottles designed for reuse.
Thermo Scientific™ Wide-Mouth HDPE Oblong with Lined Closure
Save valuable space and stabilize samples within the shipping container. Certified to meet EPA Performance Based Specifications for metals, cyanide and fluoride analyses.
Thermo Scientific™ Nalgene™ Narrow-Mouth Economy HDPE Bottles: Bulk Pack, Assembled Available on GSA/VA Contract for Federal Government customers only.
Conduct light-duty, general purpose lab applications with Thermo Scientific™ Nalgene™ Narrow-Mouth Economy HDPE Bottles, which are lightweight and break-resistant.
Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles without Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Protect light-sensitive product with these opaque amber packaging bottles, which meet requirements of light-resistant containers per USP latest edition.
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Design, assemble and certify your own combination packing by using safe and economical Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles.
Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip Available on GSA/VA Contract for Federal Government customers only.
Repeatedly dispense reagents accurately using these dropper bottles, an excellent alternative to pipetting devices.
Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Repeatedly carry out reliable routine dispensation with Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles, made of low-density polyethylene.
Thermo Scientific™ Nalgene™ LDPE Bottles with Dropper Assembly Available on GSA/VA Contract for Federal Government customers only.
Dispense one drop at a time or in a stream. These LDPE narrow-mouth bottles include a polyethylene dropping pipette and neoprene bulb inserted through the closure.
Thermo Scientific™ Nalgene™ Square Natural PPCO Graduated Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Easily measure contents with molded-in graduations on the Thermo Scientific™ Nalgene™ Square Natural PPCO Graduated Bottles with Closure.
Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles Available on GSA/VA Contract for Federal Government customers only.
Perform biopharmaceutical aseptic fluid transfer or sampling applications with the tri-clover flange of Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles.
Thermo Scientific™ Nunc™ PETG Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
Thermo Scientific™ Nalgene™ Certified Wide-Mouth Amber HDPE Bottle with Closure
Recommended for critical projects where sample containment and transport is of elevated concern, featuring durable construction that withstands the rigors of field operations.
Thermo Scientific™ Nalgene™ HDPE Biotainer™ Bottles Available on GSA/VA Contract for Federal Government customers only.
Eliminate the cost of cleaning, sterilizing and its associated validations with these certified sterile and ready-to-use bottles.
  spinner