Plastic Roller Bottles
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.
Corning™ Disposable Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polystyrene for excellent visibility and tissue culture-treated for optimal cell attachment. Corning™ Disposable Roller Bottles allow Initial scale-up of adherent cells with intermittent exposure of cells to the air and the media.
Corning™ Roller Bottle with Easy Grip Cap Available on GSA/VA Contract for Federal Government customers only.
One piece seamless construction with printed graduations
Corning™ Polystyrene Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Treated with patented Corning CellBIND™ surface treatment
Corning™ Expanded-Surface Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Features a ribbed design that doubles the growth area without increasing bottle size. Corning™ Expanded Surface Roller Bottles one-piece seamless polystyrene construction prevents warping or bowing during use.
Corning™ CellBIND™ 850cm Squared Polystyrene Roller Bottle With Easy Grip Cap Available on GSA/VA Contract for Federal Government customers only.
Features large knurls designed for ergonomic handling.
Corning™ Roux Culture Bottles
For growing mass cultures and single or monolayer cultures. When bottle is laid flat, it will hold approximately one half of the stated capacity of solution.
Thermo Scientific™ Nunc™ PETG Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
BD Falcon™ Roller Bottles
Compatible with manual and automated filling systems
  spinner