Round Glass Narrow Mouth Bottles
Qorpak™ Clear Boston Round Bottles, Vacuum and Ionized with Phenolic Pulp/Vinyl Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideally used when content integrity and visibility is important. Qorpak™ Clear Boston Round Bottles, Vacuum and Ionized with Phenolic Pulp/Vinyl Cap is cleaned using Vacuum and Ionization process.
Qorpak™ Clear Boston Round Bottles, Vacuum and Ionized with Phenolic PolyCone Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideally used when content integrity and visibility is important. Qorpak™ Clear Boston Round Bottles, Vacuum and Ionized with Phenolic PolyCone Cap is cleaned using Vacuum and Ionization process.
Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized: With Phenolic PolyCone Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized: With Phenolic PolyCone Cap ensures product integrity for long-term storage.
Qorpak™ Amber Boston Round Bottles, Ultra Clean Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles, Ultra Clean ensures product integrity for long-term storage.
Qorpak™ Clear Boston Round Bottles with Green Thermoset F217 and PTFE Caps Available on GSA/VA Contract for Federal Government customers only.
For liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with Green Thermoset F217 and PTFE Cap help to insure product integrity for long term storage.
Qorpak™ Safety Coated Amber Boston Round Bottles: With Phenolic PolyCone Caps Available on GSA/VA Contract for Federal Government customers only.
Ideal for protecting light sensitive products from UV rays and handling volatile products
Qorpak™ Clear Oval Prescription Ware Bottles
For easy product identification
Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap Available on GSA/VA Contract for Federal Government customers only.
For storing liquid samples. Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap are clear for easy viewing of contents.
Qorpak™ Clear Boston Round Bottles, Cleaned for Semi-Volatiles Available on GSA/VA Contract for Federal Government customers only.
Glass sample containers cleaned for pesticides PCB's and metals
Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized: With Thermoset F217 and PTFE Cap ensures product integrity for long-term storage.
Qorpak™ Amber Boston Round Bottles with Green Thermoset F217 and PTFE Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Green Thermoset F217 and PTFE Cap ensures product integrity for long-term storage.
Qorpak™ Clear Boston Round Bottles with Polypropylene Cap and PTFE Disc Available on GSA/VA Contract for Federal Government customers only.
For liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with Polypropylene Cap and PTFE Disc help to insure product integrity for long term storage.
Qorpak™ Clear Boston Round Bottles Without Cap Available on GSA/VA Contract for Federal Government customers only.
For liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles Without Cap help to insure product integrity for long term storage.
Fisherbrand™ Safety Coated Clear Boston Round Bottles without Cap Available on GSA/VA Contract for Federal Government customers only.
Ideal lab bottle for handling volatile substances. Fisherbrand™ Safety Coated Clear Boston Round Bottles without Cap are easier to grip and harder to break.
Qorpak™ Clear Boston Round Bottles, Ultra Clean Available on GSA/VA Contract for Federal Government customers only.
Designed for those needing containers cleaned for general lab use or fluid sampling. Qorpak™ Clear Boston Round Bottles, Ultra Clean feature a proprietary cleaning process that removes up to 99.5% of particles that are 5 microns and larger.
Fisherbrand™ Safety Coated Clear Boston Round Bottle with Black Phenolic PolyCone Cap Available on GSA/VA Contract for Federal Government customers only.
Ideal lab bottle for handling volatile substances. Fisherbrand™ Safety Coated Clear Boston Round Bottle with Black Phenolic PolyCone Cap is easier to grip and harder to break.
Qorpak™ Clear Boston Round Bottles with Black Phenolic Pulp/Tin Foil Cap Available on GSA/VA Contract for Federal Government customers only.
General-use bottles for liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with Black Phenolic Pulp/Tin Foil Cap offer maximum visibility and sample integrity.
Qorpak™ Amber Boston Round Bottles with Black Phenolic Rubber Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Black Phenolic Rubber Cap ensures product integrity for long-term storage.
Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap Available on GSA/VA Contract for Federal Government customers only.
Ideal general purpose bottle. Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap ensure integrity for long term storage.
Qorpak™ Amber Boston Round Bottles Without Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles Without Cap ensures product integrity for long-term storage.
Fisherbrand™ Clear Boston Round Bottles with PTFE-Faced PE-Lined Cap Available on GSA/VA Contract for Federal Government customers only.
For storing liquid samples. Fisherbrand™ Clear Boston Round Bottles with PTFE-Faced PE-Lined Cap are ideal for use when content integrity is important.
Kimble™ KIMAX™ Narrow-Mouth Solution Bottles with Color-Coded PTFE standard taper Flathead Stoppers
Plastic-coated for added handling protection; color-coded for easy identification
Qorpak™ Clear Boston Round Bottles with Black Phenolic PolyCone Caps Available on GSA/VA Contract for Federal Government customers only.
General-use bottles for liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with Black Phenolic PolyCone Cap offer maximum visibility and sample integrity.
Fisherbrand™ Clear Boston Round Bottles with Black Phenolic PolyCone Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles. Fisherbrand™ Clear Boston Round Bottles with Black Phenolic PolyCone Cap are ideal when content integrity is important.
Qorpak™ Clear Boston Round Bottles with White Polypropylene Hole Cap and Bonded PTFE/Silicone Septa Available on GSA/VA Contract for Federal Government customers only.
For liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with White Polypropylene Hole Cap and Bonded PTFE/Silicone Septa help to insure product integrity for long term storage.
Fisherbrand™ Amber Boston Round Bottles with Black Phenolic PolyCone Lined Caps Available on GSA/VA Contract for Federal Government customers only.
For long term storage and also offers protection for UV-sensitive contents. Fisherbrand™ Amber Boston Round Bottles with Black Phenolic PolyCone Lined Caps are ideal for use when content integrity and UV protection is important.
  spinner