Square Narrow Mouth Bottles
Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Conserve space with these square, narrow-mouth HDPE bottles. HDPE bottles are chemically and break resistant, yet are economically priced for a wide range of lab uses.
Thermo Scientific™ Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Easily measure and dispense using these translucent, square narrow-mouth PPCO bottles, featuring mouths wider than conventional narrow-mouth bottles.
Thermo Scientific™ Nalgene™ Square HDPE Graduated Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Save space with these square, translucent bottles that provide visible liquid levels and molded-in graduations for easy measuring.
Thermo Scientific™ Nalgene™ Square Natural PPCO Graduated Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Easily measure contents with molded-in graduations on the Thermo Scientific™ Nalgene™ Square Natural PPCO Graduated Bottles with Closure.
Thermo Scientific™ Nalgene™ Square Polycarbonate Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Store aqueous solutions with durable and autoclavable Thermo Scientific™ Nalgene™ Square Polycarbonate Bottles, also ideal for short-term storage of media.
Thermo Scientific™ Nalgene™ Square Polycarbonate Graduated Bottles without Closure: Tray Pack Available on GSA/VA Contract for Federal Government customers only.
These bottles are durable and autoclavable, and feature an easy-to-handle square design. They are ideal for storage of aqueous solutions and short-term storage of media.
Bel-Art™ SP Scienceware™ Buttress Cap Bottles
Excellent for sampling from tank cars, tank trucks and barges
Bel-Art™ SP Scienceware™ Book Bottles with Spigot
Features a gasketed spigot threaded into the bottle, allowing the user to dispense right from the shelf
Qorpak™ Natural HDPE Square Bottles Available on GSA/VA Contract for Federal Government customers only.
Suggested for chemical, pharmaceutical, cosmetic and food applications. Qorpak™ HDPE Square Bottles feature a space-saving design.
Bel-Art™ SP Scienceware™ Book Bottles
Polypropylene bottles stack book-like on shelves for efficient use of space
Kimble™ KIMAX™ Closure with Rubber Liner
Replacement part for Kimble 14250, 14915 and 14925
Bel-Art™ SP Scienceware™ Square Bottles
Suited for use in limited shelf space and are very good for storing or shipping samples
  spinner