Square Plastic Wide Mouth Bottles
Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Transport, sample or store liquids and dry materials with these space-efficient bottles featuring excellent chemical resistance to most acids, bases and alcohols.
Thermo Scientific™ Nalgene™ Square Wide-Mouth HDPE Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Easily fill and pour with square, wide-mouth bottles, featuring molded-in graduations and uniform walls that are resistant to splitting or puncturing.
Thermo Scientific™ Nalgene™ Square Wide-Mouth PPCO Bottles with Closure: Bulk Pack, Autoclavable Available on GSA/VA Contract for Federal Government customers only.
Save valuable storage space with easy-to-grip square, wide-mouth PPCO bottles, which feature durable, uniform walls resistant to splitting or puncturing.
Thermo Scientific™ Nalgene™ Square Wide-Mouth HDPE Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Easily fill Thermo Scientific™ Nalgene™ Square Wide-Mouth HDPE Bottles. Excellent chemical resistance makes these bottles suitable for a wide variety of uses.
Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles Available on GSA/VA Contract for Federal Government customers only.
Protect light-sensitive liquids or dry materials from exposure with these space-efficient bottles featuring sturdy walls and a wide mouth for easy filling.
Thermo Scientific™ Nalgene™ Square Wide-Mouth PPCO Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO Bottles. Contact-clear, autoclavable and resistant to acids, alcohols and bases.
Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Save space with the rectangular design of Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure, a good choice when space is at a premium.
Thermo Scientific™ Nalgene™ Rectangular Opaque Amber HDPE Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Help protect light-sensitive liquids or dry materials from exposure with Thermo Scientific™ Nalgene™ Rectangular Opaque Amber HDPE Bottles.
Thermo Scientific™ Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure: Autoclavable Available on GSA/VA Contract for Federal Government customers only.
Autoclave and store contaminated slides and other small lab equipment with the Thermo Scientific™ Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure.
Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure Available on GSA/VA Contract for Federal Government customers only.
Collect, store and transport dry chemicals and environmental samples with this large square wide-mouth HDPE bottle.
Fisherbrand™ Graduated HDPE Rectangular Bottles
Save valuable shelf space — while providing greater materials storage
Corning™ Square Polycarbonate Storage Bottles
Space-Saving square shape requires less space (13 to 20%) on the shelf or in the autoclave. Corning™ Square Polycarbonate Storage Bottles are ideal for mixing, sampling and storage.
Fisherbrand™ Square Bottom Drosophila Bottle
Wide ledge for paper lid closure
Gosselin™ Octagonal Polyethylene Terephtalate Bottles Available on GSA/VA Contract for Federal Government customers only.
Designed with a leak-resistant closure system
  spinner