Autosampler Vial Closure Accessories
Simport™ Scientific Capinsert™ For Cryovial Tubes
Inserts fit precisely into the cap of the Cryovial™ for color identification
Restek™ Dual Vespel™ Ring Inlet Seals
Extend column lifetime by preventing oxygen from reaching the column with these inlet seals. Restek™ Dual Vespel™ Ring Inlet Seals eliminate the need for a washer and ensure very little torque is needed to make a leak-tight seal.
  spinner