Autosampler Vial Septa
Thermo Scientific™ 8mm Autosampler Vial Septa Available on GSA/VA Contract for Federal Government customers only.
Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of applications.
Thermo Scientific™ 13mm Autosampler Vial Septa Available on GSA/VA Contract for Federal Government customers only.
13mm Septa for use with threaded caps Snap/Crimp Caps are available in PTFE/ Red Rubber, PTFE/Silicone or Red PTFE/White Silicone/Red PTFE.
Thermo Scientific Septa for 8-425 Screw Thread Caps
Thermo Scientific™ National Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of applications.
Thermo Scientific™ National Target™ 10mm Septa for 10-425 Threaded Caps Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Target 10mm Septa are designed for use with 10-425 threaded caps.
Thermo Scientific™ 10mm Autosampler Vial Septa Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Target 10mm Septa are designed for use with 10-425 threaded caps.
Thermo Scientific™ 9mm Autosampler Vial Septa Available on GSA/VA Contract for Federal Government customers only.
9mm Septa are available in PTFE/ Red Rubber, PTFE/Silicone or Red PTFE/White Silicone/Red PTFE to suit a variety of applications.
Thermo Scientific™ Silicone/PTFE Seals for Vial Caps Available on GSA/VA Contract for Federal Government customers only.
Compatible with Thermo Scientific Premium Vials
Thermo Scientific™ PTFE/Rubber Laminated Discs Available on GSA/VA Contract for Federal Government customers only.
For a highly inert and unreactive seal use Thermo Scientific™ PTFE/Rubber Laminated Discs.
Thermo Scientific™ PTFE/Silicone Discs Available on GSA/VA Contract for Federal Government customers only.
Combine the inertness of a PTFE coating with the resealability of silicone Thermo Scientific™ PTFE/Silicone Discs.
Thermo Scientific™ National Septa for 4mL Screw-Thread Vials Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Vial Septa are designed for use with 4mL screw-thread vials.
Thermo Scientific™ Tuf-Bond™ PTFE/Silicone Discs
Discs that combine the inertness of PTFE with the resealability of silicone
Thermo Scientific™ 20mm Septa for Headspace Vials Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National 20mm Septa for Headspace Vials are designed for use with unlined 20mm crimp seals.
Wheaton™ Septa for Open-Top Caps Available on GSA/VA Contract for Federal Government customers only.
Can be used with screw caps
Wheaton™ One–Sep Polypropylene Septas
Fits screw caps with 13–425 GPI thread
Krackeler Scientific Bleed and Temperature Optimized Septa
Maintain a leak-free seal for sample induction and exclude air from the inlet
Restek™ Merlin Microseal Septa
Parts and replacement septa
Krackeler Scientific 20mm Septa
Specifically formulated and constructed for optimum system performance
Restek™ Thermolite™ Septa
Precision molding ensures consistent, accurate fit
Krackeler Scientific Advanced Green Septa
Long-life, high-temperature septa
Krackeler Scientific General Purpose Septa
Less frequent replacement for long life time
Krackeler Scientific Merlin Microseal™ Inlet Septa
A low-bleed, longer-life alternative to standard septa for split/splitless injection and SPME
Restek™ 10mm Septa
For 2mL Big Mouth Step Vials.
Restek™ Replacement Septa
For VOA sampling vials, 60mL sample collection vials and 250mL sample collection vials
Thermo Scientific™ Hypo-Vial Septa Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Hypo-Vial Septa are ideal to use with borosilicate glass sample bottles for heating and autoclaving.
  spinner