Wash Bottles
Fisherbrand™ Labeled Wash Bottles Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Polyethylene bottles with polypropylene closures to ensure leakproof dispensing.
Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Dispense contents without tipping or shaking by using Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles, designed with a wide mouth for faster, easier filling.
Thermo Scientific™ Nalgene™ Teflon™ FEP Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Ensure contamination-free dispensing with chemical-resistant Thermo Scientific™ Nalgene™ Teflon™ FEP Solvent Wash Bottles, molded entirely of fluoropolymers.
Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Prevent separation when squeezing Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure.
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Pour without sputter with Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Wash Bottles, featuring through-the-top closure and stem molded in one piece.
Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Simplify content identification with these narrow-mouth wash bottles that feature colored closures with an integrated stem.
Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.
Thermo Scientific™ Nalgene™ Right-to-Understand Safety Wash Bottles featuring Globally Harmonized System (GHS) labeling
Prevent chemical accidents in the lab with Thermo Scientific™ Nalgene™ Right-to-Understand Wash Bottles. Bottles are labeled following the Globally Harmonized System (GHS).
Thermo Scientific™ Nalgene™ PPCO Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Reduce the risk of contamination when dispensing sterile water or reagents by using autoclavable Thermo Scientific™ Nalgene™ PPCO Wash Bottles.
Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.
Thermo Scientific™ Nalgene™ Unitary™ Red LDPE Safety Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Easily identify Thermo Scientific™ Nalgene™ Unitary™ Red LDPE Safety Wash Bottles, featuring a wide mouth for faster, safe filling.
Thermo Scientific™ Nalgene™ Color-Coded Unitary™ LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Easily fill these wide-mouth Thermo Scientific™ Nalgene™ Color-Coded Unitary LDPE Wash Bottles, which are pre-packaged, color-coded and easy to identify.
Thermo Scientific™ Nalgene™ Fluorinated HDPE Solvent Wash Bottle Available on GSA/VA Contract for Federal Government customers only.
Eliminate risk of leakage with self-venting Thermo Scientific™ Nalgene™ Fluorinated HDPE Solvent Wash Bottles, which keep the solvent in the bottle.
Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Keep liquids in clearly marked bottles that help prevent accidental misuse. Bel Art™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and “Label Your Own” bottles.
Fisherbrand™ Translucent White Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Manufactured from LDPE. Translucent White Wash Bottles deliver a single drop or fine stream.
Bel-Art™ SP Scienceware™ Right-to-Know Safety-Vented™ Wash Bottles with GHS Labeling Products for Science Education
Specially designed valve prevents pressure build-up, which eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position.
Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Are pre-printed with Chemical name and formula. Bel Art™ Solvent-Vented™ and Safety-Labeled Wash Bottles have patented valve eliminates drips and allows bottle to dispense upright or inverted.
Fisherbrand™ Safety-Labeled Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Bel Art™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information to make your workplace safer.
Justrite™ Squeeze Bottle Available on GSA/VA Contract for Federal Government customers only.
Dispense a stream of flammable liquid directly onto a small component or subassembly. The squeeze bottle and stem are made from low-density polyethylene.
Bel-Art™ SP Scienceware™ Right-to-Know, Safety-Vented Wash Bottles with GHS Labeling
Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position
Bel-Art™ SP Scienceware™ Wide Mouth Color-Code Safety Labeled Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Easy-to-read labeling printed bottle promoting safe working environment
Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
Eliminates drips and allows bottle to dispense upright or inverted, Bel Art™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles' color-coded polypropylene closures aid in quick and easy identification.
Bel-Art™ SP Scienceware™ Two-Color Wash Bottles - Safety-Vented™ and Safety Labeled, Wide-Mouth Available on GSA/VA Contract for Federal Government customers only.
A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles
Bel-Art™ SP Scienceware™ Red Wash Bottles Available on GSA/VA Contract for Federal Government customers only.
The bright red color helps identify hazardous contents at a glance
  spinner