Digital Bottletop Dispensers
Wheaton™ Socorex™ 520 Bottle-Top Dispensers Available on GSA/VA Contract for Federal Government customers only.
These accurate, user-friendly bottle-top dispensers are an affordable alternative to their all-glass counterparts.
BrandTech™ Dispensette™ S Digital Bottletop Dispensers
The Dispensette™ S Digital Bottletop Dispensers feature a “floating piston” design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.
BrandTech™ Dispensette™ S Organic Digital Bottletop Dispensers
The Dispensette™ S Digital Organic Bottletop Dispensers feature a “floating piston” design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for dispensing of organic solvents, concentrated acids and peroxides.
BrandTech™ BRAND™ Dispensette™ III, Digital Adjustable Bottle Top Dispensers
Uses a floating piston design with no seals to wear or replace, for long service life
BrandTech™ Digital Dispensette™ Organic Bottle-Top Dispensers
Ideal for HPLC and combinatorial chemistry labs
  spinner