Carboys
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Carboys with Handles Available on GSA/VA Contract for Federal Government customers only.
Store and transport solids and powders with the Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Carboys with Handles.
Thermo Scientific™ Nalgene™ Wide-Mouth Autoclavable Polypropylene Carboys with Handles Available on GSA/VA Contract for Federal Government customers only.
Easily clean and dispense with the convenient wide opening of this carboy. Wide-shoulder handles enable easy carrying and pouring, even with gloved hands.
Thermo Scientific™ Nalgene™ Rectangular Polypropylene Carboys with Closure Available on GSA/VA Contract for Federal Government customers only.
Store media, solutions and sterile water in these rectangular polypropylene carboys, featuring a space-saving design and wide mouth opening for filling and cleaning.
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboy with Spigot Available on GSA/VA Contract for Federal Government customers only.
Store and dispense solutions and media using these autoclavable polypropylene carboys, featuring a spigot boss molded into the carboy for convenience and strength.
Thermo Scientific™ Nalgene™ Round LDPE Carboys with Spigot Available on GSA/VA Contract for Federal Government customers only.
Collect and dispense distilled water, acids and reagents with Thermo Scientific™ Nalgene™ Round LDPE Carboys with Spigot.
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with 1.5 in. Sanitary Flanges Available on GSA/VA Contract for Federal Government customers only.
Use these carboys as dispensing ports or with secure hookup for supply reservoirs to systems such as fermentors or chromatography columns.
Thermo Scientific™ Nalgene™ Polypropylene Quick-Filling/Venting Closures Available on GSA/VA Contract for Federal Government customers only.
Maintain the integrity of a container and its contents with Thermo Scientific™ Nalgene™ Quick Filling/Venting Closures.
Thermo Scientific™ Nalgene™ Handle for Single-Use Carboy Available on GSA/VA Contract for Federal Government customers only.
Easily transport your single-use carboy with this stainless steel handle.
Thermo Scientific™ Nalgene™ Polypropylene Quick Action Spigots Available on GSA/VA Contract for Federal Government customers only.
Incorporate Thermo Scientific™ Nalgene™ Polypropylene Quick-Action Spigots, including an HDPE tubing adapter which accepts tubing with 1/4 or 5/16″ I.D with Carboys.
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with Bottom Tubulation Available on GSA/VA Contract for Federal Government customers only.
Ensure more complete drainage with the low outlet of these polypropylene carboys, excellent for distilled water and culture media.
Thermo Scientific™ Nalgene™ Round LDPE Carboys Available on GSA/VA Contract for Federal Government customers only.
Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles for easy carrying and pouring.
Thermo Scientific™ Nalgene™ Autoclavable Carboys Available on GSA/VA Contract for Federal Government customers only.
Choose Thermo Scientific™ Nalgene™ Autoclavable Carboys for large batches of culture media, vaccine product production, bulk media and chemical storage.
Thermo Scientific™ Nalgene™ End Caps Available on GSA/VA Contract for Federal Government customers only.
Sanitary ports for easy fill/ing and dispensing; available in polycarbonate or polypropylene.
Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers Available on GSA/VA Contract for Federal Government customers only.
Store and transport dry and solid materials, specimens and general lab supplies. These stackable containers feature positive-lock lids that form a tight seal.
Thermo Scientific™ Nalgene™ Polypropylene Sanitary Carboys with 3″ Sanitary Neck Design Available on GSA/VA Contract for Federal Government customers only.
Perform aseptic fluid transfer or sampling in biopharmaceutical applications. This carboy's sanitary design offers fewer surfaces for contamination.
Thermo Scientific™ Nalgene™ Polypropylene Heavy-Duty Vacuum Carboys Available on GSA/VA Contract for Federal Government customers only.
Ideal for the most extreme service conditions, this heavy-duty vacuum carboy is useful as a vacuum trap and will hold full vacuum up to eight hours.
Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Closure Available on GSA/VA Contract for Federal Government customers only.
Store media, solutions and sterile water in Nalgene Rectangular Clearboys. Featuring a space-saving design and extreme impact resistance. Ideal for cold room use.
Thermo Scientific™ Nalgene™ Top Works™ Flexible Systems for Nalgene Bottles, Carboys and Media Bottles Available on GSA/VA Contract for Federal Government customers only.
Create aseptic handling solutions for liquid transfer applications with Thermo Scientific™ Nalgene™ Top Works™ Flexible Systems for Nalgene Bottles, Carboys and Media.
Thermo Scientific™ Nalgene™ 13L HDPE Jerrican with Tethered Polypropylene Closure Available on GSA/VA Contract for Federal Government customers only.
Store reagents with the Thermo Scientific™ Nalgene™ Jerrican with Tethered Polypropylene Closure.
Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick Filling/Venting Closures Available on GSA/VA Contract for Federal Government customers only.
Promote faster installation using these replacement coupling inserts, which support one-handed connection and disconnection when applied to mating coupling bodies.
Thermo Scientific™ Nalgene™ Spigots for Storage Tanks Available on GSA/VA Contract for Federal Government customers only.
Ensure safe and efficient liquid dispensing with Thermo Scientific™ Nalgene™ Spigots for Storage Tanks, for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.
Thermo Scientific™ Nalgene™ HDPE Jerricans with Closure Available on GSA/VA Contract for Federal Government customers only.
Pour easily and accurately with the angled integral spout on Thermo Scientific™ Nalgene™ HDPE Jerricans.
Thermo Scientific™ Nalgene™ Fluorinated HDPE Carboys with Closure Available on GSA/VA Contract for Federal Government customers only.
Enhance long-term container performance, prevent permeation loss and yield lower extractables. A cost-effective alternative to costly fluoropolymer containers.
Thermo Scientific™ Nalgene™ Silicone Sanitary Gaskets for Nalgene™ Carboys Available on GSA/VA Contract for Federal Government customers only.
Protect against leakage, breakage and contamination with these platinum-cured silicone gaskets, ensuring the highest degree of purity for biopharmaceutical applications.
Thermo Scientific™ Nalgene™ Rectangular HDPE Carboy with Spigot Available on GSA/VA Contract for Federal Government customers only.
Store and dispense media, solutions and sterile water with space-saving Thermo Scientific™ Nalgene™ Rectangular HDPE Carboys.
Thermo Scientific™ Nalgene™ Fluorinated HDPE Jerricans with Closure Available on GSA/VA Contract for Federal Government customers only.
Enhance long-term container performance and prevent permeation loss with Thermo Scientific™ Nalgene™ Fluorinated Jerricans with Closure, intended for hard use.
Thermo Scientific™ Nalgene™ Heavy-Duty Wide-Mouth HDPE Jug with Closure Available on GSA/VA Contract for Federal Government customers only.
Store solids or powders in the Thermo Scientific™ Nalgene™ Heavy-Duty Wide-Mouth HDPE Jug with Closure, featuring the largest opening in the Nalgene carboy line.
Thermo Scientific™ Nalgene™ Amber HDPE Carboy Available on GSA/VA Contract for Federal Government customers only.
Protect photosensitive chemicals, reagents, buffers and standards when mixing and storing. These carboys meet the USP requirements for storage of light sensitive materials.
Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ with Closure Available on GSA/VA Contract for Federal Government customers only.
Conduct large-volume media and culture preparation with these polycarbonate Nalgene Clearboys, particularly when visual inspection is required.
Thermo Scientific™ Nalgene™ Rectangular HDPE Carboys without Spigots Available on GSA/VA Contract for Federal Government customers only.
Easily fill, transfer and clean using the wide mouth opening of these HDPE rectangular carboys. They feature a sturdy, space-saving design.
  spinner