Antibiotics
Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) (50mg/mL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Produced by bacterium Micromonospora rhodorangea and acts by binding ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells
Gibco™ Hygromycin B (50mg/mL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Water-soluble antibiotic purified from bacterium Streptomyces hydroscopicus
Gibco™ Sterile Puromycin Dihydrochloride Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Aminonucleoside antibiotic produced by Streptomyces alboniger
Gibco™ Penicillin-Streptomycin (10,000 U/mL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents
CAS: 69-52-3 Molecular Formula: C16H21N3NaO4S Molecular Weight (g/mol): 374.411 InChI Key: BSFVNXCYXDYHOD-ZQDFAFASSA-N Synonym: Ampicillin PubChem CID: 131673879 IUPAC Name: (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium SMILES: [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]
Gibco™ Blasticidin S HCl (10mg/mL), Sterile Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Selective antibiotic in concentration range of 50G–100μg/mL
G418 Sulfate (White to Off-white Powder), Fisher BioReagents
CAS: 108321-42-2 Molecular Formula: C20H44N4O18S2 Molecular Weight (g/mol): 692.702 InChI Key: UHEPSJJJMTWUCP-TUWLDMFGSA-N PubChem CID: 134129582 IUPAC Name: (2R,3R,4R,5R)-2-[(1S,2R,3R,4S,6R)-4,6-diamino-3-[(2S,3S,4R,5S,6R)-3-amino-4,5-dihydroxy-6-[(1R)-1-hydroxyethyl]oxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid SMILES: CC(C1C(C(C(C(O1)OC2C(CC(C(C2O)OC3C(C(C(CO3)(C)O)NC)O)N)N)N)O)O)O.OS(=O)(=O)O.OS(=O)(=O)O
Gibco™ Zeocin™ Zeocin Selection Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Selection antibiotic, formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus
Kanamycin Sulfate (White Powder), Fisher BioReagents
CAS: 25389-94-0 Molecular Formula: C18H38N4O15S Molecular Weight (g/mol): 582.575 InChI Key: OOYGSFOGFJDDHP-IZQIRFRQSA-N PubChem CID: 134129479 IUPAC Name: (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid SMILES: C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
Gibco™ Antibiotic-Antimycotic (100X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Anti-mycotic, antibiotic for prevention of cell culture contamination
Corning™ Penicillin-Streptomycin Solution Available on GSA/VA Contract for Federal Government customers only.
Effective against Gram-positive and Gram-negative bacteria. Corning™ Penicillin-Streptomycin Solution is a mix of the antibiotics Penicillin (10,000 IU) and Streptomycin (10,000µg/mL) in a 100-fold working concentration.
Blasticidin S HCl, Corning™ Available on GSA/VA Contract for Federal Government customers only.
Synonym: blasticidin s hcl
Corning™ G418 Sulfate, 50mg/mL Available on GSA/VA Contract for Federal Government customers only.
Blocks polypeptide synthesis by hindering protein elongation. Corning™ G418 Sulfate is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.
Corning™ Hygromycin B Solution Available on GSA/VA Contract for Federal Government customers only.
Blocks polypeptide synthesis and inhibits elongation.
Sulfanilamide Certified ACS, Fisher Chemical
CAS: 63-74-1 Molecular Formula: C6H8N2O2S Molecular Weight (g/mol): 172.202 MDL Number: MFCD00007939 InChI Key: FDDDEECHVMSUSB-UHFFFAOYSA-N Synonym: 4-Aminobenzenesulfonamide, P-Anilinesulfonamide PubChem CID: 5333 ChEBI: CHEBI:45373 IUPAC Name: 4-aminobenzenesulfonamide SMILES: C1=CC(=CC=C1N)S(=O)(=O)N
Picloram, 95%, Alfa Aesar™
CAS: 1918-02-1 Molecular Formula: C6H3Cl3N2O2 Molecular Weight (g/mol): 241.45 MDL Number: MFCD00012101 InChI Key: NQQVFXUMIDALNH-UHFFFAOYSA-N Synonym: picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k PubChem CID: 15965 ChEBI: CHEBI:34922 IUPAC Name: 4-amino-3,5,6-trichloropyridine-2-carboxylic acid SMILES: C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
Penicillin-Streptomycin, Dual Antibiotic Solution, MP Biomedicals
Dual antibiotic solution
Concentration: Penicillin 5,000IU/mL, Streptomycin 5mg/mL
Effective against gram-positive and gram-negative bacteria
Carbenicillin (Disodium Salt), Fisher BioReagents
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.363 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L Synonym: α-Carboxybenzylpenicillin PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)C(=O)[O-])C(=O)[O-])C.[Na+].[Na+]
Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Produced by bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells
Gibco™ Blasticidin S HCl, powder Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Bacterial selective antibiotic in concentration range of 50G–100μg/mL
Gibco™ Fungizone™ Fungizone Antimycotic Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Formulation of amphotericin B with sodium deoxycholate as an emulsifying agent to maintain the suspension
Puromycin Dihydrochloride, Fisher BioReagents
CAS: 58-58-2 Molecular Formula: C22H31Cl2N7O5 Molecular Weight (g/mol): 544.434 InChI Key: MKSVFGKWZLUTTO-ZKWLAQDLSA-N PubChem CID: 131632508 IUPAC Name: (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride SMILES: CN(C)C1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)NC(=O)C(CC4=CC=C(C=C4)OC)N)O.Cl.Cl
Gibco™ Penicillin-Streptomycin-Glutamine (100X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
for prevention from bacterial contamination in cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
Gibco™ Molecular Probes™ Gentamicin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea
Vancomycin hydrochloride, >900 microgram/mg, ACROS Organics™
CAS: 1404-93-9 Molecular Formula: C66H76Cl3N9O24 Molecular Weight (g/mol): 1485.723 InChI Key: LCTORFDMHNKUSG-WOJSORDYSA-N PubChem CID: 124080918 SMILES: CC1C(C(CC(O1)OC2C(C(C(OC2OC3=C4C=C5C=C3OC6=C(C=C(C=C6)C(C(C(=O)NC(C(=O)NC5C(=O)NC7C8=CC(=C(C=C8)O)C9=C(C=C(C=C9C(NC(=O)C(C(C1=CC(=C(O4)C=C1)Cl)O)NC7=O)C(=O)O)O)O)CC(=O)N)NC(=O)C(CC(C)C)NC)O)Cl)CO)O)O)(C)N)O.Cl
Chloramphenicol (Crystalline Powder), Fisher BioReagents
CAS: 56-75-7 Molecular Formula: C11H12Cl2N2O5 Molecular Weight (g/mol): 323.126 InChI Key: WIIZWVCIJKGZOK-RKDXNWHRSA-N Synonym: Chloromycetin, D-(?)-threo-2, 2-Dichloro-N-[?-hydroxy-?-(hydroxymethyl)-?-(4-nitrophenyl)ethyl]acetamide PubChem CID: 5959 ChEBI: CHEBI:17698 IUPAC Name: 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide SMILES: C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]
Corning™ Antibiotic-Antimycotic Solution Available on GSA/VA Contract for Federal Government customers only.
Packaged in plastic bottles
Alfa Aesar™ Vancomycin hydrochloride, Molecular Biology Grade
CAS: 1404-93-9 Molecular Formula: C66H76Cl3N9O24 Molecular Weight (g/mol): 1485.723 MDL Number: MFCD03613611 InChI Key: LCTORFDMHNKUSG-WOJSORDYSA-N PubChem CID: 124080918 SMILES: CC1C(C(CC(O1)OC2C(C(C(OC2OC3=C4C=C5C=C3OC6=C(C=C(C=C6)C(C(C(=O)NC(C(=O)NC5C(=O)NC7C8=CC(=C(C=C8)O)C9=C(C=C(C=C9C(NC(=O)C(C(C1=CC(=C(O4)C=C1)Cl)O)NC7=O)C(=O)O)O)O)CC(=O)N)NC(=O)C(CC(C)C)NC)O)Cl)CO)O)O)(C)N)O.Cl
Corning™ G418 Sulfate, Powder Available on GSA/VA Contract for Federal Government customers only.
Packaged in plastic bottles
Amphotericin B, MP Biomedicals
CAS: 1397-89-3 Molecular Formula: C47H73NO17 Molecular Weight (g/mol): 924.091 MDL Number: MFCD00877763 InChI Key: APKFDSVGJQXUKY-IYVSRKHGSA-N Synonym: Amphomoronal, Fungizone, Amphotericine B PubChem CID: 134129663 IUPAC Name: (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car SMILES: CC1C=CC=CC=CC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(CC(C(CCC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O
  spinner