Cell Culture Growth Factors and Hormones
Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, Phenol Red-Free, LDEV-Free Available on GSA/VA Contract for Federal Government customers only.
Solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma. Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, Phenol Red-Free, LDEV-Free is tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
MP Biomedicals™ Biomedicals Mycoplasma Stain Kit
Highly sensitive test detects mycoplasma and other prokaryotic organisms in under two hours
MP Biomedicals™ Mycoplasma Removal Agent (MRA)
Mycoplasma Removal Agent (MRA) is a derivative of the quinoline family of antibiotics and eliminates mycoplasma contamination in cell cultures by inhibiting mycoplasma DNA gyrase.
Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, LDEV-Free
Suited for applications requiring highly defined basement membrane preparation.
Hygiena™ Contamination Control Kits, ATP Test
High-precision test for measuring total and free ATP in liquid samples
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Cell Boost 5™ Supplement
Provides cell culture nutrients such as lipids, amino acids, vitamins, and growth factors. HyClone™ Cell Boost™ 5 Supplement is designed through Metabolic Pathway Design approach to support demand in the area of fed-batch cell culture and media customization.
  spinner