Dissociation Reagents
Corning™ Matrigel™ GFR Membrane Matrix Available on GSA/VA Contract for Federal Government customers only.
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ GFR Basement Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
Corning™ Matrigel™ Membrane Matrix Available on GSA/VA Contract for Federal Government customers only.
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
Corning™ Matrigel™ hESC-Qualified Matrix Available on GSA/VA Contract for Federal Government customers only.
Compatible with mTeSR™1-devloped by StemCell Technologies. Corning™ Matrigel™ hESC-Qualified Matrix is widely accepted as an alternative substrate for the culture of hES cells as well as human induced pluripotent stem (iPS) cells.
Gibco™ GlutaMAX™ Supplement Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
An alternative to L-glutamine, with increased stability that improves cell health
Gibco™ HEPES (1M) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A zwitterionic organic chemical buffering agent commonly used in cell culture media
Gibco™ Insulin-Transferrin-Selenium (ITS -G) (100X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A basal medium supplement to reduce the amount of fetal bovine serum (FBS) needed to culture cells
Gibco™ CTS™ TrypLE™ Select Enzyme Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
CTS™ TrypLE™ Select Enzyme
Gibco™ B-27™ Supplement (50X), minus vitamin A Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
B-27 Supplement (50X), minus vitamin A
Gibco™ N-2 Supplement (100X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
N-2 Supplement (100X)
Gibco™ L-Glutamine (200mM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ready-to-use stock solution that acts as a cell culture supplement
Gibco™ StemPro™ Accutase™ Cell Dissociation Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
StemPro Accutase Cell Dissociation Reagent
Corning™ T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Rat Available on GSA/VA Contract for Federal Government customers only.
Promotes polyclonal activation and IL-2 proliferation of most antigenically sensitized rat or mouse lymphoid T-cells
Corning™ Matrigel™ Membrane Matrix HC Available on GSA/VA Contract for Federal Government customers only.
High concentration (HC) formulation is ideal for applications in which a high protein concentration augments the growth of tumors; allows a Matrigel basement membrane matrix plug to maintain its integrity after subcutaneous injection into mice
Gibco™ B-27™ Supplement, minus insulin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
B-27 Supplement, minus insulin
Gibco™ ACK Lysing Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow
Gibco™ SynerGy™ Sterile Distilled Water Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
CAS: 7732-18-5 Molecular Formula: H2O Molecular Weight (g/mol): 18.015 InChI Key: XLYOFNOQVPJJNP-UHFFFAOYSA-N Synonym: dihydrogen oxide, dihydrogen monoxide PubChem CID: 962 ChEBI: CHEBI:15377 IUPAC Name: oxidane SMILES: O
Corning™ Cell-Tak Cell and Tissue Adhesive Available on GSA/VA Contract for Federal Government customers only.
Attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials. Corning™ Cell-Tak Cell and Tissue Adhesive is a specially formulated protein solution extracted from marine mussel, Mytilus edulis.
Gibco™ KnockOut™ Serum Replacement Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
KnockOut™ Serum Replacement
Corning™ Fibronectin, Human Available on GSA/VA Contract for Federal Government customers only.
Promotes cellular attachment, spreading, and proliferation on tissue-culture surfaces. Corning™ Fibronectin, Human can be used as an additive to serum-free culture media.
Corning™ Cell Recovery Solution
Allows for the recovery of cells cultured on BD Matrigel™ Basement Membrane Matrix for subsequent biochemical analyses
Gibco™ MEM Non-Essential Amino Acids Solution (100X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as a growth supplement for cell culture medium, to increase cell growth and viability
Gibco™ Pluronic F-68 Non-ionic Surfactant (100X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A non-ionic surfactant used to control shear forces in suspension cultures
HyClone™ Trypsin Protease
Derived from porcine pancreas. HyClone™ Trypsin Protease is used to disaggregate adherent cells from substrate or surface to which they are attached.
Corning™ Molecular Biology Reagents Available on GSA/VA Contract for Federal Government customers only.
Used in molecular biology, nucleic acid purification applications
  spinner