Extracellular Matrices
BD Annexin V Binding Buffer
BD Annexin V Binding Buffer
Formulated for use in flow cytometric assays utilizing Annexin V
Corning™ MITO+ Serum Extender
Corning™ MITO+ Serum Extender
Used to culture a variety of cells under serum-free or serum-reduced conditions
BD Leukocyte Activation Cocktail, with BD GolgiPlug™
BD Leukocyte Activation Cocktail, with BD GolgiPlug™
For elicitating primary cytokine response from T cells
MP Biomedicals™ Hormones: Dexamethasone
MP Biomedicals™ Hormones: Dexamethasone
Used to study apoptosis, cell signaling pathways and gene expression. MP Biomedicals Hormones: Dexamethasone acts as anti-inflammatory glucocorticoid that induces the production of phospholipase A2 inhibitory protein (lipocortin).
BD Recombinant Human GM-CSF
BD Recombinant Human GM-CSF
Stimulates precursor cells of granulocytes, macrophages, and eosinophils
BD Recombinant Mouse GM-CSF
BD Recombinant Mouse GM-CSF
Has potent stimulatory effects on growth and differentiation of bone marrow progenitor cells
Corning™ Bovine Pituitary Extract (BPE)
Corning™ Bovine Pituitary Extract (BPE)
Supplement used in the culture of a variety of epithelial and endothelial cells
BD Recombinant Human Interleukin-6 (IL-6)
BD Recombinant Human Interleukin-6 (IL-6)
An absorbance assay based on the Beers-Lambert law
MP Biomedicals™ Prostaglandin E2, γ-irradiated, ≥99%
MP Biomedicals™ Prostaglandin E2, γ-irradiated, ≥99%
Prostaglandin E2 is used in cell culture applications for the study of prostaglandin regulated cell signaling and gene regulation.
BD Recomninant Rat GM-CSF
BD Recomninant Rat GM-CSF
Cytokine having potent stimulatory effects on growth and differentiation of bone marrow progenitor cells
BD Recombinant Rat Interleukin-4 (IL-4)
BD Recombinant Rat Interleukin-4 (IL-4)
Potent lymphoid cell growth factor
BD Recombinant Human Interleukin-7 (IL-7)
BD Recombinant Human Interleukin-7 (IL-7)
Potent lymphoid cell growth factor which affects pre-B, pro-B, and early T cells
  spinner