Growth Factor Products
Gibco™ MarrowMAX™ Bone Marrow Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Fully-supplemented medium for short-term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures
Corning™ Epidermal Growth Factor (EGF), Mouse Natural (Receptor Grade) Available on GSA/VA Contract for Federal Government customers only.
Enhance the proliferation of a wide variety of cell types
Corning™ Epidermal Growth Factor (EGF), Human Recombinant Available on GSA/VA Contract for Federal Government customers only.
Low-molecular-weight mitogenic protein that stimulates proliferation of wide variety of cell types in vitro. Corning™ Epidermal Growth Factor (EGF), Human Recombinant growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.
Promega™ Growth Factors
Obtained from recombinant human DNA expressed in E. coli (rh), male mouse submaxillary glands (m), or natural human sources
Corning™ 7S Nerve Growth Factors (NGF), Mouse Natural
Applications include the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro.
Corning™ Insulin-like Growth Factor (IGF-I), Human Recombinant (Culture Grade)
Has mitogenic activity for several cell types, including malignant cells
Corning™ Vascular Endothelial Growth Factor (VEGF), Human Recombinant
Stimulates proliferation of cultured endothelial cells, induces migration of endothelial cells and monocytes
Corning™ Epidermal Growth Factor (EGF), Human Recombinant
Enhance the proliferation of a wide variety of cell types
Corning™ Platelet-Derived Growth Factor-BB (PDGF-BB), Human Recombinant
Designed specifically for use in basic cell biology research
Corning™ Transforming Growth Factor-b (TGF-b), Human Natural
Can stimulate or inhibit proliferation, depending on the context of its action
MP Biomedicals™ Stem Cell Factor, Human, Recombinant
Stimulates the proliferation of bone marrow cells in 5-fluorouracil treated mice
MP Biomedicals™ Other Growth Factors and Supplements
Suitable for supplementing cell cultures and investigating various metabolic processes
ZeptoMetrix Corporation Recombinant Human Growth Factors Available on GSA/VA Contract for Federal Government customers only.
Mediates the process of clonal expansion and maintenance of T-cell proliferation
  spinner