Cell Culture Dishes, Plates and Flasks
PYREX™ Watch Glasses with Fire-Polished Edges 
Molded for table stacking
PYREX™ Crystallizing Dishes 
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization
Corning™ U-Shaped Cell Culture Flasks 
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Bel-Art™ SP Scienceware™ Conway Diffusion Cells 
For Obrink method of determining carbon dioxide, ammonia or alcohol in blood or serum by diffusion method. Bel Art™ SP Scienceware™ Conway Diffusion Cells are steam autoclavable at 121°C (250°F).
Corning™ Cell Culture Treated Flasks 
Manufactured from optically clear virgin polstyrene for optimum visibility and distortion-free microscopic viewing. Corning™ Cell Culture Treated Flasks are available in a variety of sizes, designs and cap styles to meet laboratory diagnostic needs.
Corning™ HYPERFlask™ M Cell Culture Vessels 
Designed specifically for manual use, high-yield and high-performance. Specifically designed for manual use. Corning™ HYPERFlask™ M Cell Culture Vessel uses a multi-layer gas permeable growing surface for efficient gas exchange.
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
Corning™ Falcon™ Cell Culture Multi Flasks 
Grow cells faster and easier, increasing productivity in cell culture workflow. Corning™ Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
DWK Life Sciences Kimble™ KIMAX™ Crystallizing Dishes
Ideal for storage and crystallization
Falcon™ Tissue Culture Treated Flasks 
Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Corning™ Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.
Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks 
Perform large cell mass applications such as drug screening and scale up with these cell-culture treated TripleFlasks.
Thermo Scientific™ Nunc™ EasYFlask™ Cell Culture Flasks 
Enhance cell attachment, growth and differentiation with Thermo Scientific™ Nunc™ Cell Culture Treated EasYFlasks™.
Fisherbrand™ Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Perfect for cell growth, cell yields aim on little or medium input volume.
Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks 
Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.
Corning™ CELLine Disposable Bioreactor 
Cultivation flask for secreted product, recombinant protein expression and high-yield monoclonal antibody (MAb) production
Thermo Scientific™ Nunc™ Nunclon™ Δ Cell Factory Components and Replacement Parts 
Accessories for filling and emptying the cell factories with media
Fisherbrand™ PTFE Watch Glasses
Inert to all materials except molten alkali metals.
Dual Manufacturing Company Aluminium Dishes 
Heavy guage aluminum construction
Thermo Scientific™ BioLite Cell Culture Treated Flasks 
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite Cell Culture Treated Flasks, guaranteed to be sterile, non-pyrogenic and non-cytotoxic.
Corning™ U-Shaped Cell Culture Flasks 
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Saint-Gobain Chemware™ PTFE Watch Glasses
Used for weighing, crystallization, and drying
Corning™ U-Shaped Cell Culture Flasks 
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
CoorsTek™ Porcelain Combustion Boats
AD-998 High-Alumina labware
Greiner Bio-One CELLSTAR™ TC Treated Cell Culture Flasks
Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR™ TC Treated Cell Culture Flasks are free of detectable DNase, RNase and human DNA.
Corning™ Cell Culture Untreated Flasks 
Printed with lot numbers for ease in traceability
Corning™ Ultra-Low Attachment Flasks 
Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultra-Low Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Greiner Bio-One CELLSTAR™ TC Treated Lo-Profile Cell Culture Flasks
Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR™ TC Treated Lo-Profile Cell Culture Flasks is free of detectable DNase, RNase and human DNA.