Plastic Special Purpose Dishes
Bel-Art™ SP Scienceware™ Conway Diffusion Cells
For Obrink method of determining carbon dioxide, ammonia or alcohol in blood or serum by diffusion method. Bel Art™ SP Scienceware™ Conway Diffusion Cells are steam autoclavable at 121°C (250°F).
  spinner