Mammalian Cell Culture Media
Corning™ Minimum Essential Medium Eagle (Mod.)
Formulated without sodium bicarbonate to prevent clumping due to moisture absorption
Corning™ DMEM with L-Glutamine, 4.5g/L Glucose and Sodium Pyruvate Available on GSA/VA Contract for Federal Government customers only.
Subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ DMEM with L-Glutamine, 4.5g/L Glucose and Sodium Pyruvate is hybridoma tested.
Gibco™ Recovery™ Cell Culture Freezing Medium, DMSO (10%) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Complete, ready-to-use freezing medium for cryopreservation of a wide variety of mammalian cells, including CHO-S, CHO-K1, HEK 293, Jurkat, and NIH 3T3
Gibco™ DMEM, Powder, High Glucose, No Sodium Bicarbonate Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, High Glucose, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Gibco™ DMEM, High Glucose Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose is used in cell culture processing applications.
Gibco™ Expi293™ Expression Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for growth, transfection of suspension-adapted human embryonic kidney (HEK) 293 cells
Gibco™ Neurobasal™ Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used in the growth of neuronal cells from hippocampus, cortex and otregions of the brain. Gibco™ Neurobasal™ Medium is a basal medium that meets the special cell culture requirements of pre-natal and embryonic neuronal cells when used with Gibco™ B-27™ Supplement.
Gibco™ RPMI 1640 Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM, High Glucose, Pyruvate Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, Pyruvate is used in cell culture processing applications.
Gibco™ DMEM/F-12, Powder Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, Powder is used in cell culture processing applications.
Gibco™ Essential 8™ Flex Medium Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Essential 8™ Flex is a xeno-free and feeder-free medium that supports the culture of pluripotent stem cells (PSCs) without the need for weekend feeding. Gibco™ Essential 8™ Flex Medium Kit is provided in a convenient two-component kit (500mL basal medium and 10mL supplement).
Gibco™ DMEM, Powder, High Glucose, Pyruvate, No Sodium Bicarbonate Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, High Glucose, Pyruvate, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Corning™ RPMI 1640 Medium (Mod.) 1X with L-Glutamine Available on GSA/VA Contract for Federal Government customers only.
Guaranteed low endotoxin levels ≤0.025EU/mL
HyClone™ HyQ™-RS Reduced Serum Media
For applications requires lower concentration of serum supplementation
Corning™ Dulbecco's Modification of Eagle's Medium (DMEM) 1X1, Powder Available on GSA/VA Contract for Federal Government customers only.
Subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ Dulbecco's Modification of Eagle's Medium (DMEM) 1X1, Powder is hybridoma tested.
Gibco™ Opti-MEM™ I Reduced Serum Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
An improved Minimal Essential Medium that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology
Gibco™ Advanced DMEM/F-12 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ Advanced DMEMF/12 is used in cell culture processing applications.
Gibco™ DMEM/F-12, HEPES Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, HEPES is used in cell culture processing applications.
Corning™ NutriStem™ hPSC XF Medium
Supports long-term culture of human pluripotent stem cells
Corning™ DMEM With L-Glutamine and 4.5g/L Glucose; Without Sodium Pyruvate Available on GSA/VA Contract for Federal Government customers only.
Subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ DMEM With L-Glutamine and 4.5g/L Glucose; Without Sodium Pyruvate is hybridoma tested.
HyClone™ ADCF-MAb Media
Increase process yields for the manufacture of monoclonal antibodies (MAbs) with HyClone™ ADCF-MAb Media, a protein-free media containing no animal-derived components.
Gibco™ DMEM, High Glucose, GlutaMAX™ Supplement, Pyruvate Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, GlutaMAX™ Supplement, Pyruvate is used in human ex vivo tissue and cell culture processing applications.
Lonza™ BioWhittaker™ Hybridoma Serum-Free Culture Media: HL-1 Complete
Prescreened in actual hybridoma systems to ensure minimal variation
Gibco™ StemPro™-34 SFM (1X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
StemPro™-34 SFM (1X)
Gibco™ DMEM/F-12, GlutaMAX™ Supplement Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, GlutaMAX™ Supplement is used in cell culture processing applications.
Lonza™ BioWhittaker™ X-VIVO™ 15 Hematopoietic Serum-Free Culture Media
For adoptive immunotherapy and cellular therapy research. BioWhittaker™ X-VIVO™ 15 Hematopoietic Serum-Free Culture Media supports the proliferation of lymphocytes, including tumor infiltrating lymphocytes (TIL) and macrophages under serum-free conditions.
Corning™ RPMI 1640 Medium (Mod.) Available on GSA/VA Contract for Federal Government customers only.
Formulated without sodium bicarbonate to prevent clumping due to moisture absorption
Gibco™ RPMI 1640 Medium (ATCC Modification), HEPES, Liquid Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Originally developed to culture human leukemic cells in suspension and as a monolayer
  spinner