Powdered Classical Cell Culture Media
Corning™ Minimum Essential Medium Eagle (Mod.)
Formulated without sodium bicarbonate to prevent clumping due to moisture absorption
Gibco™ DMEM, Powder, High Glucose, No Sodium Bicarbonate Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, High Glucose, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Gibco™ DMEM/F-12, Powder Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, Powder is used in cell culture processing applications.
Gibco™ DMEM, Powder, High Glucose, Pyruvate, No Sodium Bicarbonate Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, High Glucose, Pyruvate, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Corning™ Dulbecco's Modification of Eagle's Medium (DMEM) 1X1, Powder Available on GSA/VA Contract for Federal Government customers only.
Subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ Dulbecco's Modification of Eagle's Medium (DMEM) 1X1, Powder is hybridoma tested.
Corning™ RPMI 1640 Medium (Mod.) Available on GSA/VA Contract for Federal Government customers only.
Formulated without sodium bicarbonate to prevent clumping due to moisture absorption
Corning™ DMEM/Ham's F-12 50/50 Mix, Powder Available on GSA/VA Contract for Federal Government customers only.
Used for culture of variety of cell types under low serum conditions. Corning™ DMEM/Ham's F-12 50/50 Mix, Powder ensures improved stability.
Gibco™ DMEM/F-12, Powder, HEPES Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, Powder, HEPES is used in cell culture processing applications.
Gibco™ DMEM, Powder, Low Glucose, Pyruvate, No Sodium Bicarbonate Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, Low Glucose, Pyruvate, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
HyClone™ DMEM, Powder with High Glucose
Supports cell culture growth and biomanufacturing processes with powdered DMEM high glucose media. HyClone™ DMEM, Powder with High Glucose is manufactured using ISO 9001 processes.
HyClone™ DMEM/HIGH Powdered Media
Supports cell culture growth and biomanufacturing processes with powdered DMEM high glucose media. HyClone™ DMEM/HIGH Powdered Media is manufactured using ISO 9001 processes.
Corning™ Ham's F-12 Medium (Mod.)
With L-Glutamine; without Sodium Bicarbonate
HyClone™ RPMI 1640 Media, Powder
Support cell culture growth and biomanufacturing processes. HyClone™ RPMI 1640 Media, Powder is manufactured using ISO 9001 certified processes.
MP Biomedicals™ Medium 199 (modified) with Hanks' Salts with Glutamine without Sodium Bicarbonate
Medium 199 (modified) with Hanks' Salts with Glutamine without Sodium Bicarbonate
MP Biomedicals™ 5-Fluoroorotic Acid [ 5-FOA] (POWDER) Monohydrate
Pale yellow crystalline powder with applications in yeast cell culture
MP Biomedicals™ Murashige and Skoog Medium, without Sucrose, Agar, IAA, and Kinetin
Murashige and Skoog Medium is widely used plant tissue culture growth medium. It has proven effective in the culture of tissue derived from moncotyledons and dicotyledons. This medium, originally formulated to support tobacco callus, has been used to support callus initiation and growth, the growth of cells in suspension culture and the regeneration of shoots and plantlets from explants.
MP Biomedicals™ Minimum Essential Medium Eagle (Modified), with Earle's Salts and L-Glutamine, without Sodium
Minimum Essential Medium Eagle (Modified), with Earle's Salts and L-Glutamine, without Sodium, Powder
MP Biomedicals™ D-(+)- RAFFINOSE PENTAHYDRATE (POWDER)
Crystalline white, pure (≥98%) raffinose pentahydrate powder with applications in the areas of yeast cell culture and gene expression
MP Biomedicals™ Glycerol, Microbiological Grade
Glycerol, Microbiological Grade
HyClone™ DMEM/F12 1:1 Media, Powder
Supports cell culture growth and biomanufacturing processes with powdered DMEM high glucose media. HyClone™ DMEM/F12 1:1 Media, Powder is manufactured using ISO 9001 processes.
MP Biomedicals™ Donor Horse Serum
Donor Horse Serum, Golden Brown Solution
MP Biomedicals™ D-(+)- GALACTOSE (POWDER) Anhydrous
White anhydrous powder with applications in the areas of yeast cell culture and gene expression
MP Biomedicals™ D-(+)- RAFFINOSE (POWDER)
Highly pure, white crystalline powder with applications in cryopreservation
RPMI 1640 Medium, Powder, MP Biomedicals
Formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent.
MP Biomedicals™ FLUCONAZOLE
Selective inhibitor of fungal cytochrome P-450 sterol C-14 α-demethylation
  spinner