Cell Culture Dishes
Corning™ TC-Treated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro
Corning™ Ultra-Low Attachment Dishes Available on GSA/VA Contract for Federal Government customers only.
Unique covalently bonded hydrogel surface is hydrophilic and neutrally charged
Falcon™ Standard Tissue Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.
Corning™ Falcon™ Tissue Culture Dish with Grid Available on GSA/VA Contract for Federal Government customers only.
Features flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning™ Falcon™ Easy-Grip Tissue Culture Dishes are uniquely designed to simplify lifting one dish or a small stack of dishes.
Thermo Scientific™ BioLite Cell Culture Treated Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite Cell Culture Treated Dishes, guaranteed to be sterile, non-pyrogenic and non-cytotoxic.
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Thermo Scientific™ Nunc™ Cell Culture/Petri Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs.
Thermo Scientific™ Nunc™ OmniTray™ Single-Well Plate Available on GSA/VA Contract for Federal Government customers only.
Screen libraries, DNA hybridization, standard petri dish techniques and bacterial growth with Thermo Scientific™ Nunc™ OmniTrays™.
Falcon™ Organ Culture Dish Available on GSA/VA Contract for Federal Government customers only.
Vacuum-gas plasma tissue culture treated polystyrene dish
Thermo Scientific™ Nunc™ Rectangular Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.
Corning™ Untreated 245mm Square BioAssay Dishes Available on GSA/VA Contract for Federal Government customers only.
Designed with a generous growth area of 500cm2 to allow for larger assays and better overall use of space in incubators. Corning™ Untreated 245mm Square Low Profile BioAssay Dishes provide a superior option for applications where cell attachment is not desired.
Corning™ Falcon™ Easy-Grip Tissue Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Features flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning™ Falcon™ Easy-Grip Tissue Culture Dishes are uniquely designed to simplify lifting one dish or a small stack of dishes.
Corning™ Primaria™ Tissue Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Modified polystyrene surface for enhanced cell culture. Corning™ Primaria™ Tissue Culture Dishes feature flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells.
Corning™ Untreated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface Available on GSA/VA Contract for Federal Government customers only.
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface.
Thermo Scientific™ Nunc™ Glass Bottom Dishes
Using Nunc Glass Bottom Dishes in your research applications gives you the flexibility to convert your culture device into an imaging device.
Thermo Scientific™ Nunc™ Non-Treated Multidishes Available on GSA/VA Contract for Federal Government customers only.
Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
Thermo Scientific™ Nunc™ MicroWell™ MiniTrays
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Corning™ BioCoat™ Fibronectin TC-treated Culture Dish
Matrix components are applied as an even, optically clear coating
Thermo Scientific™ Nunclon™ Sphera™ Dishes
Features a superior cell culture surface with demonstrated performance that elevates stem cell and cell culture research.
Corning™ CellBIND™ Surface Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically-clear virgin polystyrene for excellent visibility. Corning™ CellBIND™ Suface Culture Dishes are treated for optimal cell attachment to help cells adapt to reduced serum or serum-free media.
Corning™ BioCoat™ Poly-D-Lysine Culture Dishwith Coverslip Bottom
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ BioCoat™ Gelatin Culture Dish
Pre-coating with gelatin (obtained from porcine gelatin) eliminates time-consuming preparation
Corning™ BioCoat™ Poly-D-Lysine/Laminin Culture Dish
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Corning™ BioCoat™ Poly-L-Lysine Culture Dishes
Provide excellent substrate for cell attachment and differentiation of primary neurons, glial, neuroblastoma and glioma cells
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
MP Biomedicals™ PlantCon™ Plant Tissue Culture Container
Sterilized Base for PLANTCON™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.
  spinner