Specialty Surfaced Cellware and Inserts
Corning™ Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts Available on GSA/VA Contract for Federal Government customers only.
Featuring a microscopically transparent polyester membrane that is TC-treated for optimal cell attachment and growth. Corning™ Transwell™ Multiwell Plate with Polyester Membrane Inserts provide better visibility under phase contrast microscopy and allow assessment of cell viability and monolayer formation.
MilliporeSigma™ Millicell™ Culture Plate Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed for improved cell differentiation and expanded applications involving analytical cell biology. MilliporeSigma™ Millicell™ Culture Plate Inserts incorporate PTFE Biopore™ membranes with 0.4μm pores. Ready-to-use 30 or 12mm inserts for 6 and 24-well plates.
Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning™ Matrigel Matrix Available on GSA/VA Contract for Federal Government customers only.
Provide rapid and reproducible quantification of cell invasion in vitro. Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning Matrigel Matrix™ cell culture inserts are pre-coated with extracellular matrix proteings for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and differentiation.
Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts Available on GSA/VA Contract for Federal Government customers only.
Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.
Corning™ Snapwell™ Culture Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed with a ring supported membrane for use with diffusion or Ussing chambers once cells are grown to confluence. Corning™ Snapwell™ Culture Inserts are primarily used for transport and electrophysiological studies.
MilliporeSigma™ Millicell™ Culture Plate Inserts: PCF Available on GSA/VA Contract for Federal Government customers only.
Designed for improved cell differentiation and expanded applications involving analytical cell biology
Falcon™ Cell Culture Inserts Available on GSA/VA Contract for Federal Government customers only.
Use with 24-well plates. Corning™ Falcon™ Cell Culture Inserts (cell culture on permeable supports) results in greater morphological and/or physiological differentiation of many cell types.
Corning™ BioCoat™ Matrigel™ Invasion Chamber: With GFR Matrigel Matrix Available on GSA/VA Contract for Federal Government customers only.
Provide rapid and reproducible quantification of cell invasion in vitro
MilliporeSigma™ Millicell™ Cell Culture Insert Plates Available on GSA/VA Contract for Federal Government customers only.
Designed to support cell growth and differentiation of endothelial and epithelial cell lines
Corning™ BioCoat™ Poly-D-Lysine/Laminin Culture Slide Available on GSA/VA Contract for Federal Government customers only.
Tested for the ability to promote neurite outgrowth with NG108 rat glioma/mouse neuroblastoma cells
Falcon™ Cell Culture Inserts: For Use With 6-Well Plates Available on GSA/VA Contract for Federal Government customers only.
Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types
Corning™ BioCoat™ Control Insert
Use alongside BD BioCoat™ inserts when studying effects of the ECM component present on the BD BioCoat cell culture insert
Falcon™ Cell Culture Inserts: For Use With 12-Well Plates Available on GSA/VA Contract for Federal Government customers only.
Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types
Corning™ Costar™ Transwell™-COL Collagen-Treated Coated Membrane Insert Available on GSA/VA Contract for Federal Government customers only.
Transparent, collagen-treated microporous membranes promote cell attachment and spreading
Corning™ BioCoat™ Poly-D-Lysine Glass Multiwell Culture Slide Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ BioCoat™ Collagen I Glass Multiwell Culture Slide Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Thermo Scientific™ Nunc™ Polycarbonate Cell Culture Inserts in Multi-Well Plates Available on GSA/VA Contract for Federal Government customers only.
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Corning™ BioCoat™ Tumor Invasion Multiwell Plate Available on GSA/VA Contract for Federal Government customers only.
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
Corning™ BioCoat™ Fibronectin, Human Cell Culture Insert
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
Corning™ BioCoat™ Fibronectin Culture Slide, Human
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Research tool for transport, co-culture, migration and invasion studies. Greriner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion through physical surface (TC) treatment.
MilliporeSigma™ Millicell™ Culture Plate Inserts: HATF
Designed for improved cell differentiation and expanded applications involving analytical cell biology
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation
Evaluate and quantitate endothelial cell invasion using real-time fluorescence and non-destructive sample preparation
Corning™ BioCoat™ Collagen I Inserts with PET Membrane
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ BioCoat™ TC-treated Multiwell Variety Packs
Choose from different groups of cellware conveniently packaged together in popular combinations
Stemgent™ Alvetex™ 3D Cell Growth Well Inserts
Multiple formats for versatility in cell culturing
Corning™ Falcon™ Not TC-Treated In Vitro Fertilization (IVF)/Low Wall Dish
Pre-tested plasticware for in vitro fertilization (IVF)
  spinner