Equine Sera
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Donor Equine Serum (U.S.)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care.
Corning™ Donor Horse Serum, U.S. Sourced Available on GSA/VA Contract for Federal Government customers only.
Horse serum is often used in testing media for mycoplasma contamination or as a blocking agent and controls in immunoassays. It can also be combined with fetal bovine serum for the culture of rat adrenal pheochromocytoma (PC12) cells.
MP Biomedicals™ Whole Serum, Horse, Purified, 2mL
Whole Horse Serum with buffer salts
  spinner