Cellular Imaging Instrumentation Accessories
Invitrogen™ Coverglass Removal Tool Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Minimize the risk of breakage of chambered coverglass during cell culture and fluorescence imaging applications. CultureWell&Trade; Coverglass Removal Tool enables the chambered upper structure to be easily separated from the coverglass without the need for excessive force.
Objective Lenses for ArrayScan™ and CellInsight™ High-Content Platforms Available on GSA/VA Contract for Federal Government customers only.
Expand the functionality of your Thermo Scientific™ high-content instrument
  spinner