Centrifuge Tubes and Bottles
Fisherbrand™ Universal Test Tube Plug Caps
For plastic or glass test tubes.
Fisherbrand™ Colored Screw Caps
For use with nonsterile skirted- and conical-bottom tubes.
DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes, 10mL Available on GSA/VA Contract for Federal Government customers only.
Use these tubes for centrifugation up to 2980 RCF. KIMAX™ Conical-Bottom Glass Centrifuge Tubes are plastic shrink-wrapped in modular trays for cleanliness and safety. Manufactured from 51 expansion, low extractable borosilicate glass.
Fisherbrand™ Flat and Domed 8-Cap Strips, Flat Cap Strips Available on GSA/VA Contract for Federal Government customers only.
Suitable for use with 96-well PCR plates and PCR tube strips
Fisherbrand™ Premium Microcentrifuge Tubes: 1.5mL Edge
Available in eight colors or in assorted-color packs.
Falcon™ Sterile Polyethylene Snap Caps Available on GSA/VA Contract for Federal Government customers only.
For Falcon Round-Bottom Tubes
Falcon 50mL Conical Centrifuge Tubes Edge Available on GSA/VA Contract for Federal Government customers only.
Tested to withstand centrifugation of 16,000 RCF in a fully supported rotor with room temperature water. Corning™ Falcon 50mL Conical Centrifuge Tubes are ideal for cell centrifugation; pelleting; separation by density gradients.
Fisherbrand™ Sterile Microcentrifuge Tubes with Screw Caps
Choose skirted or conical-bottom style
Falcon™ 15 mL Conical Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Tested to withstand centrifugation of 12,000 RCF (polypropylene) or 1,800 RCF (polystyrene) in a fully supported rotor with room temperature water. Corning™ Falcon™ 15mL Conical Centrifuge Tubes are ideal for cell centrifugation; pelleting; separation by density gradients.
Thermo Scientific™ Screw Cap Microcentrifuge Tube Caps
Prevent leakage under demanding conditions. These screw-cap tube caps with o-rings are optimized for secure short-term storage in low temperatures.
Axygen™ MaxyClear Snaplock Microtubes, 1.7 mL Edge Available on GSA/VA Contract for Federal Government customers only.
Ensure an efficient seal that stays closed during centrifugation. Axygen™ MaxyClear Snaplock Microtubes, 1.7mL provide excellent visibility and feature a piercing port for syringe access. Maximum Recovery™ and sterile options available.
Thermo Scientific™ Capitol Vial 10mL and 12mL Threaded Transport Tubes with Pre-Assembled Screw-Caps
Fisherbrand™ Pre-Capped 10mL and 12mL Threaded Transport Tubes are fabricated from FDA approved clarified polypropylene resin for visual clarity.
Fisherbrand™ Snap Caps for Nonsterile Plastic Tubes
Closures for plastic culture tubes.
DWK Life Sciences Kimble™ Kim-Kap™ Disposable Closures Edge Available on GSA/VA Contract for Federal Government customers only.
Available in natural (clear) or choice of four colors
Corning™ Disposable Conical-Bottom Glass Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Nonsterile tubes, constructed of Pyrex™ borosilicate glass.
Fisherbrand™ Premium Microcentrifuge Tubes: 2.0mL Edge
Flat-top snap caps provide a safe, liquid-tight, reliable seal even with prolonged boiling
Thermo Scientific™ Nunc™ 15mL and 50mL Conical Sterile Polypropylene Centrifuge Tubes Edge  Available on GSA/VA Contract for Federal Government customers only.
These premium, high-quality conical tubes are environmentally friendly, and allow for increased traceability with the largest writing area on the market.
Thermo Scientific™ Molecular BioProducts™ Flange Caps
Nonautoclavable polyethylene, for use with 12 and 13mm test and culture tubes
Axygen™ MaxyClear Snaplock Microtubes, 1.5 mL Available on GSA/VA Contract for Federal Government customers only.
Ensure an efficient seal that stays closed during centrifugation. Axygen™ MaxyClear Snaplock Microtubes, 1.5mL provide excellent visibility and feature a piercing port for syringe access. Available in a wide variety of colors. Maxymum Recovery™ and sterile options available.
Thermo Scientific™ Screw Cap Micro Tubes
Pure Protection: Prepare, transfer and store samples securely with Thermo Scientific Screw Cap Microtubes. Available in multiple volumes, packaging options and clean claims.
Fisherbrand™ Screw Caps for Disposable Glass Tubes Edge Available on GSA/VA Contract for Federal Government customers only.
Phenolic caps have cemented-in rubber liners
Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes Edge
Improve sample recovery without additives or coatings. These tubes offer maximum sample recovery by significantly reducing surface binding.
Corning™ Polyethylene Caps for Cluster Tube System Edge Available on GSA/VA Contract for Federal Government customers only.
Designed to fit 1.2mL cluster tubes and cluster tube strips. Corning™ Polyethylene Caps for Cluster Tube System are used with 98-well cluster tube systems.
Axygen™ Screw Cap Transport Tubes Available on GSA/VA Contract for Federal Government customers only.
Tube volumes from 5 to 10mL
  spinner