Glass Centrifuge Tubes
DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes, 10mL Available on GSA/VA Contract for Federal Government customers only.
Use these tubes for centrifugation up to 2980 RCF. KIMAX™ Conical-Bottom Glass Centrifuge Tubes are plastic shrink-wrapped in modular trays for cleanliness and safety. Manufactured from 51 expansion, low extractable borosilicate glass.
Corning™ Disposable Conical-Bottom Glass Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Nonsterile tubes, constructed of Pyrex™ borosilicate glass.
DWK Life Sciences Kimble™ KIMAX™ 100mL 8 in. Oil Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
KIMAX™ tube used in the determination of water and sediment in crude mineral oils, fuel oils and petroleum products; of precipitate formed by centrifuging definite quantities of steam cylinder stocks, black oils and lubricating oils; and in testing for acidity of distillation residues or hydrocarbon liquids of gasoline/petroleum solvents.
DWK Life Sciences Kimble™ Conical-Bottom Glass Centrifuge Tubes, 50mL, Screw Thread Available on GSA/VA Contract for Federal Government customers only.
With a bullet-nosed shape, these disposable screw thread Centrifuge Tubes withstand centrifugation up to 2980 RCF. Closures are not supplied (for closures, see Mfr. No. 73800 and 73802).
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Reusable High Strength Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Chemically strengthened to achieve a greater mechanical strength
PYREX™ Pear-Shaped Glass Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
For use in IEC™ HN-SII Centrifuge from Thermo Scientific with 05-152-1 rotor
DWK Life Sciences Kimble™ KIMAX™ Heavy-Duty Graduated Centrifuge Tubes with Screw Caps Available on GSA/VA Contract for Federal Government customers only.
This Standard Taper KIMAX™ tube is a heavy-duty version of 45166.
DWK Life Sciences Kimble™ KIMAX™ 50mL Conical-Bottom Glass Centrifuge Tubes with Screw Thread Caps Available on GSA/VA Contract for Federal Government customers only.
This Standard Taper KIMAX™ tube is graduated and reusable, and comes with a 24-410 GPI Finish screw thread cap.
DWK Life Sciences Kimble™ KIMAX™ 50mL Heavy Duty Graduated Centrifuge Tubes with PTFE Stopper
This Standard Taper KIMAX™ tube is made with heavier-walled tubing to withstand higher centrifugation speeds. Graduated scale and legend are permanently stained on the outer surface.
DWK Life Sciences Kimble™ KIMAX™ 100mL 6 in. Short Cone Oil Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
KIMAX™ tube designed for field use in testing petroleum. Centrifuge tubes with 6-inch short cone.
Fisherbrand™ Graduated Centrifuge Tube with Screw Thread Cap Available on GSA/VA Contract for Federal Government customers only.
Reusable large capacity 100mL tube with screw thread cap and PTFE-faced rubber liner.
DWK Life Sciences Kimble™ KIMAX™ 100mL 8 in. Oil and Weathering (End Point Index) Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Can be used in the determination of residues in Liquified Petroleum (LP) gases, ASTM Method D2158.
DWK Life Sciences Kimble™ KIMAX™ 100mL Pear-Shaped Glass Centrifuge Tubes with Pennyhead Stoppers Available on GSA/VA Contract for Federal Government customers only.
KIMAX™ tube used for the determination of small quantities of solids in large volumes of liquids.  Recommended for the determination of free water and sediment in diesel and other distillate fuels, as a pass-fail indication of product quality (ASTM D2709).
DWK Life Sciences Kimble™ KIMAX™ 50mL Graduated Dairyware Glass Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Used in the determination of the solubility index of dry milk solids or whey products as specified by the American Dairy Products Institute. Also used in pulp determination of citrus products.
DWK Life Sciences Kimble™ Plain Reusable Centrifuge Tube with Screw Caps
With PTFE/rubber lined black phenolic screw cap
  spinner