Plastic Centrifuge Tubes
Falcon™ 50mL Conical Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Tested to withstand centrifugation of 16,000 RCF in a fully supported rotor with room temperature water. Corning™ Falcon™ 50mL Conical Centrifuge Tubes are ideal for cell centrifugation; pelleting; separation by density gradients.
Corning™ Centrifuge Tubes with CentriStar™ Cap Available on GSA/VA Contract for Federal Government customers only.
Features CentriStar™ caps with an easy-on/easy-off flat top and offers advanced ergonomics with wider knurls and roll-over edge design for easier gripping. Corning™Centrifuge Tubes with CentriStar™ Caps leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.
Corning™ Polypropylene Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from clear polypropylene for superior chemical resistance. Corning™ Polypropylene Centrifuge Tubes are an ideal solution for centrifuge needs.
Falcon™ 15mL Conical Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Tested to withstand centrifugation of 12,000 RCF (polypropylene) or 1,800 RCF (polystyrene) in a fully supported rotor with room temperature water. Corning™ Falcon™ 15mL Conical Centrifuge Tubes are ideal for cell centrifugation; pelleting; separation by density gradients.
Fisherbrand™ Easy Reader™ Conical Polypropylene Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Imprinted clear black graduations ensure accurate readings
Thermo Scientific™ Nunc™ 15mL and 50mL Conical Sterile Polypropylene Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
These premium, high-quality conical tubes are environmentally friendly, and allow for increased traceability with the largest writing area on the market.
Fisherbrand™ Higher-Speed 50mL Easy Reader™ Plastic Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Withstand higher speeds than other Easy Reader tubes — up to 15,000xg
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
View contents of these contact-clear, Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes easily.
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Open and close Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes with one hand due to a proprietary hinged-cap system.
Greiner Bio-One Leucosep™ Centrifuge Tubes with Porous Barrier
Efficient separation of lymphocytes and mononuclear cells.
Fisherbrand™ 50mL Graduated Polypropylene Centrifuge Tube
Skirt extends beyond the bottom of the tube so it can stand without support
Basix™ Polypropylene Centrifuge Tubes
Sterile tubes with graduated divisions
Corning™ Mini Bioreactor Centrifuge Tube
Ideal for high throughput process optimization for suspension cell culture
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Perform a plethora of applications thanks to the excellent mechanical strength of Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes.
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap Available on GSA/VA Contract for Federal Government customers only.
Easily view the contents of contact-clear, Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Caps.
Falcon™ Polypropylene Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polypropylene for high chemical and thermal resistance. Corning™ Falcon™ Polypropylene Centrifuge Tubes are tested to withstand 7500 x g centrifugation.
Eppendorf™ Eppendorf Tubes™ Conical Tubes Available on GSA/VA Contract for Federal Government customers only.
Use these tubes for the simple and safe processing of sample volumes up to 5.0mL. Eppendorf Tubes™ represent the perfect option for working with medium–sized sample volumes.
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.
Corning™ Polyethylene Terephthalate (PET) Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from crystal clear rigid PET for optimum visibility. Corning™ Polyethylene Terephthalate (PET) Centrifuge Tubes have conical bottoms and high-density polyethylene plug seal caps providing a premium centrifuge tube.
Greiner Bio-One Leucosep™ Polypropylene Round Bottom Centrifugation Tubes
For efficient separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow.
Thermo Scientific™ Nunc™ 200mL Centrifuge Tube Available on GSA/VA Contract for Federal Government customers only.
Process large batches more easily with Nunc 200mL centrifuge tubes, which are made from high-purity resins and can resist forces up to 7000 RCF.
Corning™ Polyetherimide Centrifuge Tube Cushions Available on GSA/VA Contract for Federal Government customers only.
Tube cushion provides necessary support during centrifugation preventing large capacity tubes from collapsing
Thermo Scientific™ Nalgene™ High-Speed Polycarbonate Round Bottom Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Repeatedly centrifugalize samples successfully thanks to the outstanding mechanical strenghth and high optical clarity of these polycarbonate round-bottom centrifuge tubes.
Greiner Bio-One Conical Bottom Polypropylene Test Tubes with Screw Cap
For centrifugation and applications that require chemical and thermo resistant vessels.
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap Available on GSA/VA Contract for Federal Government customers only.
Easily view contents using these strong, transparent polycarbonate centrifuge tubes, available with sealing caps for use up to 50,000 x g.
Thermo Scientific™ Nalgene™ Oak Ridge Polysulfone Centrifuge Tubes w/Screw Caps Available on GSA/VA Contract for Federal Government customers only.
Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge Polysulfone Centrifuge Tubes, available with sealing caps.
Thermo Scientific™ Nalgene™ PC Conical-Bottom Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Perform who cell separations with the help of Thermo Scientific™ Nalgene™ PC Conical-Bottom Centrifuge Tubes
Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Simplify measurements using Nalgene PPCO Conical-Bottom Centrifuge Tubes which feature molded-in graduations. Translucent PPCO has excellent chemical resistance.
  spinner