Benzoic Acid
Benzoic Acid (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Benzoic Acid CAS: 65-85-0 Purity Grade: Certified ACS Physical Form: Solid Assay: 99.5% min. Assay Percent Range: ≥99.5 % Molecular Formula: C7H6O2 Linear Formula: C6H5COOH Formula Weight: 122.12g/mol MDL Number: MFCD00002398 Synonym: Benzenecarboxylic Acid
Benzoic Acid (Crystalline/USP/FCC), Fisher Chemical
Chemical Name or Material: Benzoic Acid CAS: 65-85-0 Purity Grade: USP/FCC Physical Form: Solid Assay: 99.5 to 100.5% (FCC) Molecular Formula: C7H6O2 Linear Formula: C6H5COOH Formula Weight: 122.12g/mol MDL Number: MFCD00002398 Synonym: Benzenecarboxylic Acid
Benzoic Acid (Crystalline/Primary Standard), Fisher Chemical
Chemical Name or Material: Benzoic Acid Name Note: Primary standard for calorimetry CAS: 65-85-0 Purity Grade: Primary Standard Physical Form: Solid Molecular Formula: C7H6O2 Linear Formula: C6H5COOH Formula Weight: 122.12g/mol MDL Number: MFCD00002398 Synonym: Benzenecarboxylic Acid
Benzoic acid, 99.5%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Benzoic acid Name Note: For analysis CAS: 65-85-0 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: 99.5% min. Assay Percent Range: 99.5% Molecular Formula: C7H6O2 Linear Formula: C6H5CO2H Formula Weight: 122.12g/mol MDL Number: MFCD00002398
Benzoic Acid, Sodium Salt, 99+%, ACROS Organics™
Chemical Name or Material: Sodium benzoate CAS: 532-32-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra pure Assay: 99.0% min. (on dried substance) Assay Percent Range: 99+% Molecular Formula: C7H5NaO2 Linear Formula: C6H5CO2Na Formula Weight: 144.11g/mol MDL Number: MFCD00012463 Synonym: Benzoic acid, sodium salt
Benzoic Acid Standard, For Elementary Analysis, EMD Millipore™
Chemical Name or Material: Benzoic Acid Standard CAS: 65-85-0 Molecular Formula: C6H5COOH Formula Weight: 122.12g/mol. (molar mass) Synonym: Benzene carboxylic acid Synonym: Benzene carboxylic acid
Benzoic Acid Benzyl Ester, 99.6%, MP Biomedicals™
Chemical Name or Material: Benzyl Benzoate CAS: 120-51-4 Physical Form: Liquid Assay Percent Range: 99.6% Molecular Formula: C14H12O2 Formula Weight: 212.2g/mol Synonym: Benzoic Acid Benzyl Ester
Benzoic acid, ACS Reagent Grade, ≥99.5%, MP Biomedicals
Chemical Name or Material: Benzoic acid Name Note: For ACS analysis CAS: 65-85-0 Purity Grade: ACS Reagent Physical Form: White powder Assay Percent Range: ≥99.5% Molecular Formula: C7H6O2 Formula Weight: 122.12g/mol MDL Number: MFCD00002398
Benzoic Acid Ammonium Salt 99%, ACROS Organics™
Chemical Name or Material: Benzoic acid ammonium salt Name Note: 99% CAS: 1863-63-4 CAS Min %: 98.5 CAS Max %: 100.0 Assay: 98.5% min. Molecular Formula: C7H9NO2 Linear Formula: CO2NH4C6H5 Formula Weight: 139.15g/mol Synonym: Ammonium benzoate
Benzoic Acid Copper(II) Salt Hydrate 96%, ACROS Organics™
Chemical Name or Material: Benzoic acid copper(II) salt hydrate Name Note: 96% CAS: 66019-07-6 CAS Min %: 95.0 CAS Max %: 100.0 Assay: 95% min. Assay Percent Range: 96% Molecular Formula: C14H10CuO4·xH2O Linear Formula: (C6H5CO2)2Cu·xH2O Formula Weight: 305.77g/mol MDL Number: MFCD03066124
Benzoic Acid, BP, Ph Eur, USP grade, 99.5 to 100.5%, EMPROVE, EMD Millipore
Chemical Name or Material: Benzoic Acid Name Note: EMPROVE™ CAS: 65-85-0 Purity Grade: BP/EP/USP Physical Form: Powder Assay Percent Range: 99.5 to 100.5% Molecular Formula: C6H5COOH Linear Formula: C6H5COOH Formula Weight: 122.12g/mol Synonym: Phenylformic Acid
Benzoic Acid Products for Science Education
Chemical Name or Material: Benzoic Acid CAS: 65-85-0 Purity Grade: Laboratory Assay Percent Range: >99% Linear Formula: C6H5COOH Formula Weight: 122.12g/mol Synonym: Benzenecarboxylic Acid
  spinner