Cacodylic Acid
Cacodylic acid, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cacodylic acid Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C2H7AsO2 Linear Formula: (CH3)2As(O)OH Formula Weight: 138.01 MDL Number: MFCD00002095
Cacodylic acid, sodium salt trihydrate, 98%, pure, ACROS Organics™
Chemical Name or Material: Cacodylic acid, sodium salt trihydrate Name Note: 98% CAS: 6131-99-3 Purity Grade: Pure Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C2H6AsNaO2·3H2O Linear Formula: (CH3)2As(O)ONa·3H2O Formula Weight: 214.03 MDL Number: MFCD00149079 Synonym: Dimethylarsinic acid sodium salt
Cacodylic Acid (Crystalline), Fisher BioReagents
Chemical Name or Material: Cacodylic Acid CAS: 75-60-5 CAS Min %: 99 CAS Max %: 100 Physical Form: Powder/Solid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C2H7AsO2 Linear Formula: (CH3)2As(O)OH Formula Weight: 138.01 Synonym: Dimethylarsinic Acid
  spinner