Citric Acid
Citric Acid Anhydrous (Crystalline/USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid, Anhydrous CAS: 77-92-9 Purity Grade: USP Physical Form: Solid Assay: 99.5 to 100.5% (USP) Molecular Formula: C6H8O7 Formula Weight: 192.13g/mol MDL Number: MFCD00011669
Citric Acid Monohydrate (Granular/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Monohydrate CAS: 5949-29-1 Purity Grade: Certified ACS Physical Form: Solid Assay: 99.0 to 102.0% Assay Percent Range: 99.0 to 102.0 % Molecular Formula: C6H8O7 · H2O Linear Formula: (HO2CCH2)2C(OH)(CO2H)·H2O Formula Weight: 210.14g/mol MDL Number: MFCD00149972
Citric Acid Monohydrate (Granular/USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Monohydrate CAS: 5949-29-1 Purity Grade: USP Physical Form: Solid Assay: 99.5 to 100.5% (USP) Molecular Formula: C6H8O7 · H2O Formula Weight: 210.14g/mol MDL Number: MFCD00149972
Citric Acid Anhydrous (Crystalline/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Anhydrous CAS: 77-92-9 Purity Grade: Certified ACS Physical Form: Solid Assay: 99.5% min. Assay Percent Range: ≥99.5 % Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13g/mol MDL Number: MFCD00011669
Citric Acid Monohydrate (Crystalline/USP/EP/BP/JP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Monohydrate CAS: 5949-29-1 Purity Grade: USP/EP/BP/JP Physical Form: Solid Molecular Formula: C6H8O7 · H2O Formula Weight: 210.14g/mol MDL Number: MFCD00008765
Citric Acid U.S.P. Granular, Macron Fine Chemicals™
Chemical Name or Material: Citric Acid Name Note: Monohydrate CAS: 5949-29-1 Purity Grade: USP Physical Form: Granular Molecular Formula: C6H8O7·H2O Formula Weight: 210.14g/mol
Citric acid, trisodium salt 98%, anhydrous, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, trisodium salt CAS: 68-04-2 Purity Grade: Pure Assay: 97.5% min. Assay Percent Range: 98% Molecular Formula: C6H5Na3O7 Formula Weight: 258.07g/mol MDL Number: MFCD00012462 Synonym: Trisodium citrate
Citric acid monohydrate, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: Citric acid monohydrate CAS: 5949-29-1 Purity Grade: For analysis Assay: 99.4% min. Assay Percent Range: 99.5% Molecular Formula: C6H8O7·H2O Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H·H2O Formula Weight: 210.15g/mol MDL Number: MFCD00149972
Citric Acid, Monohydrate Granular/ACS), Macron Fine Chemicals™
Chemical Name or Material: Citric Acid, Monohydrate CAS: 5949-29-1 Purity Grade: ACS Grade Physical Form: Granular Molecular Formula: C6H8O7·H2O Formula Weight: 210.14g/mol
Citric Acid, Anhydrous, 99%, Pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid Name Note: Anhydrous CAS: 77-92-9 Purity Grade: Pure Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13g/mol MDL Number: MFCD00011669 Synonym: 2-Hydroxy-1, 2, 3-propanetricarboxylic acid
Citric Acid, 10% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Citric Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 77-92-9 CAS Min %: 9.0 CAS Max %: 11.0 Physical Form: Clear colorless liquid
Citric Acid, Granular, Multi-Compendial, USP, J.T.Baker™
Chemical Name or Material: Citric Acid CAS: 5949-29-1 Purity Grade: Multi-Compendial, USP Molecular Formula: HOC(COOH)(CH2COOH)2·H2O Formula Weight: 210.14g/mol
Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, trisodium salt hydrate Name Note: For molecular biology CAS: 114456-61-0 Purity Grade: For molecular biology Assay: 99.75% min. (on dry substance) Assay Percent Range: 99.8% Molecular Formula: C6H5Na3O7·xH2O Formula Weight: 258.08g/mol MDL Number: MFCD00150032
Citric Acid, Anhydrous p.a., ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid CAS: 77-92-9 Purity Grade: For analysis Assay: 99.5% min. Assay Percent Range: 99.5% Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13g/mol Synonym: 2-Hydroxy-1, 2, 3-propanetricarboxylic acid
Citric Acid Anhydrous ACS, Ricca Chemical
Chemical Name or Material: Citric Acid Anhydrous CAS: 77-92-9 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Physical Form: White granules
Citric Acid Solution, 50%, Biotech Reagent, J.T.Baker™
Chemical Name or Material: Citric Acid Solution Purity Grade: Biotech Reagent Molecular Formula: HOC(COOH)(CH2COOH)2 Formula Weight: 192.12g/mol
Citric acid, trisodium salt dihydrate, 99%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, trisodium salt dihydrate CAS: 6132-04-3 Purity Grade: Pure Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C6H5Na3O7·2H2O Linear Formula: NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O Formula Weight: 294.09g/mol MDL Number: MFCD00150031 Synonym: Sodium citrate tribasic dihydrate
Citric acid, trisodium salt dihydrate, 99%, for biochemistry, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, trisodium salt dihydrate CAS: 6132-04-3 Purity Grade: For biochemistry Assay Percent Range: 99% Molecular Formula: C6H5Na3O7·2H2O Linear Formula: NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O Formula Weight: 294.09g/mol Synonym: Sodium citrate tribasic dihydrate
Citric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Citric Acid CAS: 5949-29-1 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: HOC(COOH)(CH2COOH)2·H2O Formula Weight: 210.14g/mol
Citric Acid, Crystalline Powder, U.S.P. - F.C.C., J.T.Baker™
Chemical Name or Material: Citric Acid CAS: 5949-29-1 Purity Grade: USP, FCC Molecular Formula: HOC(COOH)(CH2COOH)2·H2O Formula Weight: 210.14g/mol
Citric Acid, 1% (w/w), Ricca Chemical
Chemical Name or Material: Citric Acid CAS: 77-92-9 CAS: 7732-18-5
Citric Acid, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Citric Acid CAS: 5949-29-1 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: HOC(COOH)(CH2COOH)2·H2O Formula Weight: 210.14g/mol
Citric Acid, Powder, U.S.P., J.T.Baker™
Chemical Name or Material: Citric Acid CAS: 77-92-9 Purity Grade: USP Molecular Formula: HOC(COOH)(CH2COOH)2 Formula Weight: 192.12g/mol
Citric acid, triammonium salt, +97%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, triammonium salt CAS: 3458-72-8 Purity Grade: Extra pure Assay: 97.0% min. Assay Percent Range: 97+% Molecular Formula: C6H17N3O7 Formula Weight: 243.22g/mol MDL Number: MFCD00036406 Synonym: Triammoniumcitrate
Citric Acid, U.S.P. - F.C.C., J.T.Baker™
Chemical Name or Material: Citric Acid CAS: 5949-29-1 Purity Grade: USP, FCC Molecular Formula: HOC(COOH)(CH2COOH)2·H2O Formula Weight: 210.14g/mol
Citric Acid, Powder, Multi-Compendial, J.T.Baker™
Chemical Name or Material: Citric Acid CAS: 77-92-9 Purity Grade: Multi-Compendial Molecular Formula: HOC(COOH)(CH2COOH)2 Formula Weight: 192.12g/mol
Citric Acid, Anhydrous, Reagent ACS 99.5%, ACROS Organics™
Chemical Name or Material: Citric acid CAS: 77-92-9 Purity Grade: ACS Reagent Assay: 99.5% min. Assay Percent Range: 99.6% Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13g/mol Synonym: 2-Hydroxy-1, 2, 3-propanetricarboxylic acid
Citric acid, diammonium salt, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, diammonium salt CAS: 3012-65-5 Purity Grade: Extra pure Assay: 97.5% min. Assay Percent Range: 98% Molecular Formula: C6H14N2O7 Formula Weight: 226.19g/mol MDL Number: MFCD00013068 Synonym: Diammoniumcitrate
Citric Acid Anhydrous (Crystalline), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Anhydrous CAS: 77-92-9 Physical Form: Solid Assay: 99.5% min. Assay Percent Range: ≥99.5 % Molecular Formula: C6H8O7 Formula Weight: 192.13g/mol
Citric Acid, 1.00 Molar, Ricca Chemical
Chemical Name or Material: Citric Acid CAS: 7732-18-5 CAS Min %: 82.0 CAS Max %: 86.0 CAS: 77-92-9 CAS Min %: 15.8 CAS Max %: 16.45 Physical Form: Clear colorless liquid
  spinner