Decanoic Acid
Decanoic acid, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Decanoic acid Name Note: 99+% CAS: 334-48-5 Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C10H20O2 Linear Formula: CH3(CH2)8CO2H Formula Weight: 172.27g/mol MDL Number: MFCD00004441 Synonym: Capric acid
Decanoic acid, 99%
Chemical Name or Material: Decanoic Acid CAS: 334-48-5 Molecular Formula: CH3(CH2)8CO2H Formula Weight: 172.27g/mol MDL Number: MFCD00004441 Beilstein: 1754556 Merck Index: 14,1758
2-Decenoic Acid, MP Biomedicals
Chemical Name or Material: 2-Decenoic Acid CAS: 3913-85-7 Molecular Formula: C10H18O2 Formula Weight: 170g/mol
  spinner