Formic Acid
Fisher chemical Formic Acid, 99.5+%, Optima™ LC/MS Grade Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formic Acid Name Note: 99.5% CAS: 64-18-6 Purity Grade: Optima LC/MS Assay: 99.5% min. Assay Percent Range: ≥99.5 % Molecular Formula: CH2O2 Linear Formula: HCOOH Formula Weight: 46.02g/mol Synonym: FA
Formic Acid, 88% (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formic Acid, 88% CAS: 64-18-6 Purity Grade: Laboratory Molecular Formula: CH2O2 Linear Formula: HCOOH Formula Weight: 46.03g/mol MDL Number: MFCD00003297
Formic Acid, 88% (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formic Acid, 88% CAS: 64-18-6 Purity Grade: Certified ACS Molecular Formula: CH2O2 Linear Formula: HCOOH Formula Weight: 46.03g/mol MDL Number: MFCD00003297 Synonym: Methanoic Acid
Formic Acid, Reagent ACS 88+%, ACROS Organics™
Chemical Name or Material: Formic acid CAS: 64-18-6 Purity Grade: ACS Reagent Assay: 88.0% min. Assay Percent Range: 88% Molecular Formula: CH2O2 Linear Formula: HCO2H Formula Weight: 46.02g/mol
Formic Acid, 88%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Formic Acid CAS: 64-18-6 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: HCOOH Formula Weight: 46.03g/mol
Formic acid, 97%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formic acid Name Note: 96+% CAS: 64-18-6 Purity Grade: ACS Reagent Assay: 96.0% min. Assay Percent Range: 97% Molecular Formula: CH2O2 Linear Formula: HCO2H Formula Weight: 46.02g/mol MDL Number: MFCD00003297
Formic Acid, 25% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Formic Acid Name Note: Aqueous Solution CAS: 64-18-6 CAS: 7732-18-5 Physical Form: Clear colorless liquid Assay Percent Range: 25% (v/v)
Formic Acid, 90%, BAKER ANALYZED™ Reagent, J.T.Baker™
Chemical Name or Material: Formic Acid CAS: 64-18-6 Name Note: Polymer Characterization Solvent Purity Grade: BAKER ANALYZED™ Reagent Molecular Formula: HCOOH Formula Weight: 46.03g/mol
Formic Acid, 88%, AR, For ACS analysis, Macron Fine Chemicals™
Chemical Name or Material: Formic Acid Name Note: ACS CAS: 64-18-6 Purity Grade: ACS Grade Physical Form: Liquid Molecular Formula: HCOOH Formula Weight: 46.03g/mol
Formic acid, sodium salt, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium formate Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: CHNaO2 Linear Formula: HCO2Na Formula Weight: 68.01g/mol MDL Number: MFCD00013101 Synonym: Formic acid, sodium salt
Formic acid, 99%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formic acid CAS: 64-18-6 Purity Grade: For analysis Assay: 99.0% min. Assay Percent Range: 99% Molecular Formula: CH2O2 Linear Formula: HCO2H Formula Weight: 46.02g/mol MDL Number: MFCD00003297
Formic Acid, 40% (w/w) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Formic Acid CAS: 7732-18-5 CAS Min %: 59.0 CAS Max %: 61.0 CAS: 64-18-6 CAS Min %: 39.0 CAS Max %: 41.0 Physical Form: Clear colorless liquid
Formic acid, 98+%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formic acid CAS: 64-18-6 Purity Grade: Pure Assay: 98.0% min. Assay Percent Range: 98+% Molecular Formula: CH2O2 Linear Formula: HCO2H Formula Weight: 46.02g/mol
Formic Acid (Aldehyde-Free/Sequencing), Fisher BioReagents™
Chemical Name or Material: Formic Acid Name Note: Aldehyde-free CAS: 64-18-6 CAS: 7732-18-5 Purity Grade: Sequencing Assay: 88.0% min. Assay Percent Range: ≥88.0 % Molecular Formula: CH2O2 Linear Formula: HCOOH Formula Weight: 46.02g/mol
Formic Acid, 2% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Formic Acid CAS: 7732-18-5 CAS Min %: 96.0 CAS Max %: 100.0 CAS: 64-18-6 CAS Min %: 2.16 CAS Max %: 2.25 Physical Form: Clear colorless liquid
Formic Acid, 98% (ACS), GR, EMD Millipore
Chemical Name or Material: Formic Acid, 98% (ACS) CAS: 64-18-6 Purity Grade: ACS Guaranteed Reagent
Formic Acid ACS, 98%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formic Acid CAS: 64-18-6 Purity Grade: ACS Grade, USP, NF Physical Form: Liquid Assay Percent Range: 98% Linear Formula: HCOOH Formula Weight: 46.3g/mol Synonym: Aminic Acid
Formic Acid ACS, 88%, EMD Chemicals
Chemical Name or Material: Formic Acid CAS: 64-18-6 Purity Grade: ACS Grade, USP, NF Physical Form: Liquid Assay Percent Range: 88% Linear Formula: HCOOH Formula Weight: 46.3g/mol Synonym: Aminic Acid
Formic Acid, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formic Acid CAS: 64-18-6 Purity Grade: Technical Physical Form: Liquid Assay Percent Range: 90% Linear Formula: HCOOH Formula Weight: 46.3g/mol Synonym: Aminic Acid
Formic Acid 98-100%, Suprapur™, EMD Millipore
Name Note: Suprapur™ CAS: 64-18-6 Purity Grade: Suprapur Physical Form: Clear Colorless Liquid Assay Percent Range: 98 to 100% Molecular Formula: CH2O2 Linear Formula: HCOOH Formula Weight: 46.3g/mol Synonym: Formylic Acid
Formic acid, ACS, 96+%
Chemical Name or Material: Formic Acid CAS: 64-18-6 Name Note: For analysis ACS Purity Grade: ACS Grade Molecular Formula: CH2O2 Linear Formula: HCOOH Formula Weight: 46.03g/mol MDL Number: MFCD00003297 UN Number: UN1779 Beilstein: 1209246 Merck Index: 14,4241
Formic Acid, MP Biomedicals
Chemical Name or Material: Formic Acid CAS: 64-18-6 Formula Weight: 46.02g/mol
Formic acid, ACS, 88+%
Chemical Name or Material: Formic Acid CAS: 64-18-6 Purity Grade: ACS Grade Molecular Formula: HCOOH Linear Formula: HCOOH Formula Weight: 46.03g/mol MDL Number: MFCD00003297 UN Number: 1779 Beilstein: 1209246 Merck Index: 14,4241
Formic Acid, free acid, >97%, MP Biomedicals™
Chemical Name or Material: Formic Acid, free acid CAS: 64-18-6 Physical Form: Clear colorless liquid Assay Percent Range: >97% Molecular Formula: CH2O2 Formula Weight: 46.03g/mol Synonym: Acide formique
Formic Acid ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Formic Acid CAS: 64-18-6 Purity Grade: ACS Grade Assay Percent Range: 88% Molecular Formula: HCOOH Formula Weight: 46.03g/mol
Formic Acid, Ammonium Salt, BAKER™, J.T.Baker™
Chemical Name or Material: Formic Acid, Ammonium Salt CAS: 540-69-2 Purity Grade: BAKER™ Molecular Formula: HCOONH4 Formula Weight: 63.06g/mol
Formic Acid, 88% Products for Science Education
Chemical Name or Material: Formic Acid, 88% CAS: 64-18-6 CAS: 7732-18-5 Purity Grade: Reagent Assay Percent Range: 88% w/v Linear Formula: HCOOH Formula Weight: 46.03g/mol Synonym: Hydrogen Carboxylic Acid
  spinner