Glycolic Acid
Glycolic acid, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Glycolic acid Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C2H4O3 Linear Formula: HOCH2CO2H Formula Weight: 76.04 Synonym: Hydroxyacetic Acid
Glycolic acid, 98%
Chemical Name or Material: Glycolic Acid CAS: 79-14-1 Molecular Formula: HOCH2CO2H Linear Formula: HOCH2CO2H Formula Weight: 76.05 MDL Number: MFCD00004312 UN Number: UN3261 Beilstein: 1209322 Merck Index: 14,4498
Glycolic Acid (White Crystalline Solid/Certified), Fisher Chemical
Chemical Name or Material: Glycolic Acid CAS: 79-14-1 Purity Grade: Certified Grade Physical Form: Solid Assay: ≥98.5% Assay Percent Range: ≥98.5 % Molecular Formula: C2H4O3 Linear Formula: HOCH2COOH Formula Weight: 76.05 MDL Number: MFCD00004312 Synonym: Hydroxyacetic Acid
Glycolic Acid, BAKER™, J.T.Baker™
Chemical Name or Material: Glycolic Acid CAS: 79-14-1 Molecular Formula: HOCH2COOH Formula Weight: 76.05
  spinner