Hydrofluoric Acid
Fisher chemical Hydrofluoric Acid 47-51%, Optima™, for Ultra Trace Elemental Analysis Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrofluoric Acid CAS: 7664-39-3 Purity Grade: Optima™ Molecular Formula: HF Formula Weight: 20.01g/mol MDL Number: MFCD00011346
Hydrofluoric Acid, 48-50% (Technical), Fisher Chemical
Chemical Name or Material: Hydrofluoric Acid, 48-50% CAS: 7732-18-5, 7664-39-3 CAS: 7664-39-3 Purity Grade: Technical Assay: 48 to 50 w/w% Assay Percent Range: 48 to 50 w/w % Molecular Formula: HF Formula Weight: 20.01g/mol MDL Number: MFCD00011346
Hydrofluoric Acid (TraceMetal™ Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrofluoric Acid CAS: 7664-39-3 Purity Grade: TraceMetal™ Molecular Formula: HF Formula Weight: 20.01g/mol MDL Number: MFCD00011346
Hydrofluoric Acid (Certified ACS), Fisher Chemical
Chemical Name or Material: Hydrofluoric Acid CAS: 7732-18-5 CAS: 7664-39-3 Purity Grade: Certified ACS Assay: 48.0 to 51.0 w/w% Assay Percent Range: 48.0 to 51.0 w/w % Molecular Formula: HF Formula Weight: 20.01g/mol MDL Number: MFCD00011346 Synonym: Fluorhydric Acid
Hydrofluoric Acid, ULTREX™ II Ultrapure Reagent, J.T.Baker™
Chemical Name or Material: Hydrofluoric Acid CAS: 7664-39-3 Purity Grade: ULTREX™ II Ultrapure Reagent Molecular Formula: HF Formula Weight: 20.01g/mol
Hydrofluoric acid, for analysis, 48 to 51% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrofluoric acid Name Note: p.a. CAS: 7664-39-3 CAS: 7732-18-5 Purity Grade: For analysis Assay: 48 to 51% Molecular Formula: HF Linear Formula: HF Formula Weight: 20.00g/mol MDL Number: MFCD00011346
Hydrofluoric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™
Chemical Name or Material: Hydrofluoric Acid CAS: 7664-39-3 Purity Grade: BAKER INSTRA-ANALYZED™ Plus Molecular Formula: HF Formula Weight: 20.01g/mol
Hydrogen fluoride-pyridine, 65-70%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrogen fluoride-pyridine Name Note: 65-70% CAS: 32001-55-1 Assay: 65 to 70% (HF) Assay Percent Range: 65-70% Molecular Formula: C5H6FN Formula Weight: 99.11g/mol MDL Number: MFCD00012436 Synonym: Olah';s reagent
Hydrofluoric acid, J.T.Baker™
Chemical Name or Material: Hydrofluoric acid CAS: 7664-39-3 Physical Form: Liquid Molecular Formula: HF Formula Weight: 36.46g/mol Synonym: Fluorhydric acid
Hydrofluoric acid, ACS reagent, 48-51% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrofluoric acid Name Note: 48.0-51.0% CAS: 7664-39-3 CAS: 7732-18-5 Purity Grade: ACS Reagent Assay: 48.0 to 51.0% Molecular Formula: HF Linear Formula: HF Formula Weight: 20.00g/mol MDL Number: MFCD00011346
Hydrofluoric Acid, 2% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Hydrofluoric Acid CAS: 7732-18-5 CAS Min %: 96.0 CAS Max %: 100.0 CAS: 7664-39-3 CAS Min %: 2.22 CAS Max %: 2.31 Physical Form: Clear colorless liquid
Hydrofluoric Acid ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Hydrofluoric Acid CAS: 7664-39-3 Purity Grade: ACS Assay Percent Range: 48% Molecular Formula: HF Formula Weight: 20.01g/mol
Hydrofluoric acid, J.T.Baker™
Chemical Name or Material: Hydrofluoric Acid CAS: 7664-39-3 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: HF Formula Weight: 36.46g/mol Synonym: Fluoric Acid; Fluorhydric Acid
Hydrofluoric Acid , Macron Fine Chemicals™
Chemical Name or Material: Hydrofluoric Acid Assay Percent Range: 52% Molecular Formula: HF Formula Weight: 20.01g/mol
Hydrofluoric Acid, Baker Instra-Analyzed Reagent, USP/NF Grade, For ACS analysis, J.T.Baker™
CAS: 7664-39-3 Purity Grade: USP, NF Purity Grade Notes: For ACS analysis Physical Form: Liquid Assay Percent Range: 48.0 to 51.0% Molecular Formula: HF Formula Weight: 20.01g/mol
hydrogen fluoride, pure, 37 wt.% solution in triethylamine, ACROS Organics™
Chemical Name or Material: Hydrogen fluoride CAS: 7664-39-3 CAS: 121-44-8 Purity Grade: Pure Assay: 37% (HF) Molecular Formula: HF·1/3C6H15N Linear Formula: (C2H5)3N·3HF Formula Weight: 161.21g/mol MDL Number: MFCD00011346
Hydrofluoric Acid, 1% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Hydrofluoric Acid CAS: 7732-18-5 CAS Min %: 96.0 CAS Max %: 100.0 CAS: 7664-39-3 CAS Min %: 1.92 CAS Max %: 2.0 Physical Form: Clear colorless liquid
Hydrofluoric Acid, 48%, AR, For ACS analysis, Macron Fine Chemicals™
Chemical Name or Material: Hydrofluoric Acid Name Note: ACS CAS: 7664-39-3 Physical Form: Colorless corrosive fuming solution Assay Percent Range: 48.75 to 49.2 % Molecular Formula: HF Formula Weight: 20.01g/mol
Hydrofluoric Acid, 0.4% (v/v), Ricca Chemical
Chemical Name or Material: Hydrofluoric Acid CAS: 7664-39-3 CAS: 7732-18-5 Physical Form: Clear colorless liquid Assay Percent Range: 0.4% (v/v)
Hydrofluoric Acid ACS, EMD Millipore
Chemical Name or Material: Hydrofluoric Acid Purity Grade: ACS Grade, USP, NF Physical Form: Liquid Assay Percent Range: 48% Linear Formula: HF Formula Weight: 2.1g/mol Synonym: Hydrogen Fluoride
Hydrofluoric acid, 47-51%, OmniTrace Ultra, EMD Millipore
Chemical Name or Material: Hydrofluoric acid Name Note: OmniTrace™ Ultra CAS: 7664-39-3 Purity Grade: ACS Grade, USP, NF Physical Form: Liquid Assay Percent Range: 47 to 51% Molecular Formula: HF Linear Formula: HF Formula Weight: 2.1g/mol Synonym: Hydrogen Fluoride
Hydrochloric acid, ACS, HCl 36.5-38.0%, Alfa Aesar™
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 Purity Grade: ACS Physical Form: Clear colorless liquid Assay Percent Range: 36.5-38.0% Molecular Formula: HCl Formula Weight: 36.46g/mol MDL Number: MFCD00011324 Synonym: Muriatic acid
Hydrofluoric Acid, 49%, CMOS, J.T.Baker™
Chemical Name or Material: Hydrofluoric Acid Name Note: CMOS CAS: 7664-39-3 Assay Percent Range: 49%
Hydrofluoric acid, Environmental Grade, 47-51%, Alfa Aesar™
Chemical Name or Material: Hydrofluoric acid Name Note: Single sub-boiling quartz distillation CAS: 7664-39-3 CAS: 7732-18-5 Purity Grade: Environmental Grade Physical Form: Clear colorless liquid Assay Percent Range: 47-51% Molecular Formula: HF Formula Weight: 20.01g/mol MDL Number: MFCD00011346
Hydrofluoric acid, ACS, 48-51%, Alfa Aesar™
Chemical Name or Material: Hydrofluoric acid Name Note: Special handling precautions required CAS: 7664-39-3 Purity Grade: ACS Physical Form: Liquid Assay Percent Range: 48-51% Molecular Formula: HF Formula Weight: 20.01g/mol MDL Number: MFCD00011346
  spinner