Iodic Acid
Iodic acid, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: Iodic acid Name Note: p.a. CAS: 7782-68-5 Purity Grade: For analysis Assay: ≥99.4% Assay Percent Range: 99.5% Molecular Formula: HIO3 Linear Formula: HIO3 Formula Weight: 175.91 MDL Number: MFCD00011348
Iodic Acid, ACS, 99.5%, Alfa Aesar
Chemical Name or Material: Iodic Acid CAS: 7782-68-5 Purity Grade: ACS Grade Physical Form: Powder/Chunks Assay Percent Range: 99.5% Molecular Formula: HIO3 Formula Weight: 175.91 MDL Number: MFCD00011348
Iodic acid, 99.6%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Iodic acid Name Note: For analysis ACS CAS: 7782-68-5 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: HIO3 Linear Formula: HIO3 Formula Weight: 175.91 MDL Number: MFCD00011348
Iodic Acid (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Iodic Acid CAS: 7782-68-5 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: HIO3 Formula Weight: 175.91 MDL Number: MFCD00011348 Synonym: Hydrogen Iodate
  spinner