Mandelic Acid
DL-Mandelic acid, 99+%, ACROS Organics™
Chemical Name or Material: DL-Mandelic acid Name Note: 99+% CAS: 90-64-2 Assay: 99.0 % min. (Acid) Assay Percent Range: 99+% Molecular Formula: C8H8O3 Linear Formula: C6H5CH(OH)CO2H Formula Weight: 152.15g/mol Synonym: alpha-Hydroxyphenylacetic acid
(R)-(-)-Mandelic Acid 99%, ACROS Organics™
Chemical Name or Material: (R)-(-)-Mandelic acid Name Note: 99% CAS: 611-71-2 CAS Min %: 98.5 CAS Max %: 100.0 Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C8H8O3 Linear Formula: C6H5CH(OH)CO2H Formula Weight: 152.15g/mol MDL Number: MFCD00064251 Synonym: (R)-(-)-α-Hydroxyphenylacetic acid
(S)-(+)-Mandelic Acid ≥99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: (S)-(+)-Mandelic acid Name Note: 99+% CAS: 17199-29-0 CAS Min %: 99.0 CAS Max %: 100.0 Assay: 99.0 % min. (Acid) Assay Percent Range: 99+% Molecular Formula: C8H8O3 Linear Formula: C6H5CH(OH)CO2H Formula Weight: 152.15g/mol MDL Number: MFCD00004495 Synonym: (S)-(+)-α-Hydroxyphenylacetic acid
L-Mandelic Acid, MP Biomedicals
Chemical Name or Material: L-Mandelic Acid CAS: 611-71-2 Formula Weight: 152.15g/mol
(±)-Mandelic Acid, 99%, Alfa Aesar™
Chemical Name or Material: (±)-Mandelic Acid Assay Percent Range: 99% Molecular Formula: C8H8O3 Formula Weight: 152.15g/mol
D-(-)-mandelic acid, 99%, MP Biomedicals™
Chemical Name or Material: D-(-)-mandelic acid CAS: 611-71-2 Purity Grade: 0.99 Physical Form: White crystalline flakes Assay Percent Range: 99% Molecular Formula: C8H8O3 Formula Weight: 152.1g/mol Synonym: Hydroxyphenylacetic acid
  spinner